Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900017370.mm.1 |
TC0900002104.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017370.mm.1 |
NM_001081121.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017370.mm.1 |
NM_001081121.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017366.mm.1 |
TC0900002104.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017366.mm.1 |
NM_001081121.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017366.mm.1 |
NM_001081121.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017368.mm.1 |
TC0900002104.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017368.mm.1 |
NM_001081121.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017368.mm.1 |
NM_001081121.1 |
Refseq |