Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01170633_m1
          See other 5730403I07RIK GE Assays ›
          Gene Symbol:
          5730403I07Rik
          Gene Name:
          RIKEN cDNA 5730403I07 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 77366412 - 77399416 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01170633_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5730403I07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040378.1 3 - 4 322 85
          GenBank mRNA AK017485.1 - 3 - 4 324 85
          AK161639.1 - 3 - 4 220 85
          BC048919.1 - 3 - 4 222 85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BQ715174.1 - 3 - 4 221 Chr. 9 - 77367445 - 77399315 [-] on Build GRCm38 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream muscular LMNA-interacting protein Mlip 2310046A06Rik,
          AI931782,
          CIP
          -18.5kb Sense Chr. 9 - 77102084 - 77347870 on NCBI Build GRCm38
          Downstream leucine rich repeat containing 1 Lrrc1 A430093J20Rik,
          AU016091,
          D030059I21,
          mKIAA4018
          46.6kb Sense Chr. 9 - 77430823 - 77544852 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002701.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334738 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286275 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 840145 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 231374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337519 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 32076 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130099 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080095 BC048919.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06851218 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130099 - -
          Whole Mouse Genome G4122A A_52_P668090 BC048919.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P668090 AA426741.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P668090 AK017485.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140853 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07482079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07843002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07482074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08012554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07707368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08183362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07917491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08133288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07774534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07774531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07709577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07843001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07696209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07696217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07868010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26654114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08133286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653935 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430396_PM_at - -
          Mouse Genome 430 2.0 Array 1430396_at - -
          Mouse Genome 430 2.0 Array 1430396_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70487_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70487_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900022313.mm.1 TC0900002701.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900022313.mm.1 NR_040378.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022313.mm.1 NR_040378.1 Refseq
          Mouse Expression Set 430 1430396_at - -
          Mouse Expression Set 430 1430396_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430396_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430396_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 716347 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245939 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0