Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01171220_m1
          See other 1600029O15RIK GE Assays ›
          Gene Symbol:
          1600029O15Rik
          Gene Name:
          ribosomal protein L17 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 58202897 - 58208808 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01171220_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1600029O15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033522.1 2 - 3 392 67
          GenBank mRNA AK005564.2 - 2 - 3 392 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream immunoglobulin superfamily containing leucine-rich repeat Islr AW553146 -44.2kb Anti-sense Chr. 9 - 58156246 - 58159239 on NCBI Build GRCm38
          Downstream promyelocytic leukemia Pml 1200009E24Rik,
          AI661194,
          Trim19
          8.4kb Anti-sense Chr. 9 - 58217179 - 58249796 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000728.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK005564.2 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28109 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826916 AF104414.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826916 AK038612.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826916 NM_010690.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826916 XM_618738.3 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826916 XM_906240.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361034 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102754 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984152 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102754 - -
          Whole Mouse Genome G4122A A_51_P142206 NR_033522.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P142206 AK005564.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131531 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102754 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10585824 - -
          Affymetrix HT MG-430 PM Array Plate 1454055_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10585823 - -
          Mouse Genome 430 2.0 Array 1454055_PM_at - -
          Mouse Genome 430 2.0 Array 1454055_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 665268_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 665268_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900005379.mm.1 TC0900000728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900005379.mm.1 NR_033522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900005376.mm.1 TC0900000728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900005376.mm.1 NR_033522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900005377.mm.1 TC0900000728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900005377.mm.1 NR_033522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900005378.mm.1 TC0900000728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900005378.mm.1 NR_033522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900005375.mm.1 TC0900000728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900005375.mm.1 NR_033522.1 Refseq
          Mouse Expression Set 430 1454055_PM_at - -
          Mouse Expression Set 430 1454055_at - -
          Affymetrix HT Mouse Genome 430B Array 1454055_at - -
          Affymetrix HT Mouse Genome 430B Array 1454055_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 876704 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0