Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01171347_m1
          See other KRTDAP GE Assays ›
          Gene Symbol:
          Krtdap
          Gene Name:
          keratinocyte differentiation associated protein
          Gene Aliases:
          Kdap, sk221
          Chromosome Location:
          Chr.7: 30787905 - 30791083 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01171347_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krtdap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033131.3 NP_001028303.1 4 - 5 236 115
          GenBank mRNA AK131603.1 - 4 - 5 237 115
          BC132510.1 - 4 - 5 236 40130787 115
          BC132512.1 - 4 - 5 236 40130789 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000576.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          epidermis development
          cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146223 NM_001033131.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146223 AK131603.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146223 AK131606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146223 BC132510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146223 BC132512.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146223 NM_001033131.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146223 AK131603.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146223 AK131606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146223 BC132510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146223 BC132512.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952897 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146223 NM_001033131.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146223 AK131603.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146223 AK131606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146223 BC132510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146223 BC132512.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146223 NM_001033131.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146223 AK131603.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146223 AK131606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146223 BC132510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146223 BC132512.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01493658 - -
          Whole Mouse Genome G4122A A_51_P146223 NM_001033131.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146223 AK131603.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146223 AK131606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146223 BC132510.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146223 BC132512.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146223 NM_001033131.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146223 AK131603.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146223 AK131606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146223 BC132510.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146223 BC132512.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P702887 BC145437.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06634696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05954957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06524432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06645268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06082030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06522589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06369593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06633526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06645263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06524426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06183527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06183526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06069123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06183521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06524427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05954954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05954959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06263704 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552069 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552066 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552068 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552067 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552070 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552065 - -
          Affymetrix HT MG-430 PM Array Plate 1434227_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552064 - -
          Mouse Genome 430 2.0 Array 1434227_at - -
          Mouse Genome 430 2.0 Array 1434227_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64661_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64661_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700005198.mm.1 TC0700000576.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005198.mm.1 NM_001033131.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005198.mm.1 NM_001033131.3 Refseq
          Mouse Expression Set 430 1434227_PM_at - -
          Mouse Expression Set 430 1434227_at - -
          Murine Genome U74v2 Set 99654_PM_s_at - -
          Murine Genome U74v2 Set 99654_s_at - -
          Mouse Genome 430A 2.0 Array 1434227_at - -
          Mouse Genome 430A 2.0 Array 1434227_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434227_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434227_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 819086 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220605 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2868152 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2868152 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0