Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01171350_m1
          See other RAB11A GE Assays ›
          Gene Symbol:
          Rab11a
          Gene Name:
          RAB11A, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 64715300 - 64737756 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01171350_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab11a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017382.5 NP_059078.2 4 - 5 645 133
          GenBank mRNA AB232605.1 - 4 - 5 512 133
          AK014268.1 - 4 - 5 646 133
          AK031143.1 - 4 - 5 548 133
          AK132072.1 - 4 - 5 629 133
          AK132159.1 - 4 - 5 631 133
          AK147122.1 - 4 - 5 646 133
          BC010722.1 - 4 - 5 634 2811973 133
          D50500.1 - 2 - 3 153 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002495.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab11a

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokinesis
          transport
          intracellular protein transport
          exocytosis
          cell cycle
          mitotic metaphase plate congression
          small GTPase mediated signal transduction
          metabolic process
          positive regulation of epithelial cell migration
          regulation of multivesicular body size
          positive regulation of G2/M transition of mitotic cell cycle
          protein transport
          vesicle-mediated transport
          astral microtubule organization
          neuron projection development
          melanosome transport
          Rab protein signal transduction
          multivesicular body assembly
          positive regulation of axon extension
          regulation of long-term neuronal synaptic plasticity
          regulation of protein transport
          establishment of vesicle localization
          regulation of vesicle-mediated transport
          establishment of protein localization to organelle
          protein localization to plasma membrane
          establishment of protein localization to membrane
          mitotic spindle assembly
          exosomal secretion
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          microtubule binding
          myosin V binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P444162 BC010722.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P444162 AK132072.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P444162 AK132159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P444162 AK147122.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P444162 NM_017382.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P444162 BC010722.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P444162 AK132072.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P444162 AK132159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P444162 AK147122.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P444162 NM_017382.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690156 AK157988.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690156 BC019711.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690156 NM_145379.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 179466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99994 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111317 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128632 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137760 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P444162 BC010722.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P444162 AK132072.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P444162 AK132159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P444162 AK147122.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P444162 NM_017382.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P444162 BC010722.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P444162 AK132072.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P444162 AK132159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P444162 AK147122.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P444162 NM_017382.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277505 AK132159.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277505 NM_017382.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277505 AK014268.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277505 AK031143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P444162 BC010722.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P444162 AK132072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P444162 AK132159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P444162 AK147122.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P444162 NM_017382.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P444162 BC010722.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P444162 AK132072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P444162 AK132159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P444162 AK147122.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P444162 NM_017382.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P389310 AK132159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P389310 AK147122.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P389310 NM_017382.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P389310 AK031143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P277505 AK132159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P277505 NM_017382.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P277505 AK014268.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P277505 AK031143.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123359 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07993345 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07943153 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07943151 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07993344 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07993335 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10594482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594489 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594487 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594485 - -
          Affymetrix HT MG-430 PM Array Plate 1434549_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1449256_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10594480 - -
          Mouse Genome 430 2.0 Array 1449256_a_at - -
          Mouse Genome 430 2.0 Array 1434549_at - -
          Mouse Genome 430 2.0 Array 1434549_PM_at - -
          Mouse Genome 430 2.0 Array 1449256_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 53869_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 53869_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900020946.mm.1 TC0900002495.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020946.mm.1 NM_017382.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020946.mm.1 NM_017382.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020951.mm.1 TC0900002495.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020951.mm.1 NM_017382.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020951.mm.1 NM_017382.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020948.mm.1 TC0900002495.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020948.mm.1 NM_017382.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020948.mm.1 NM_017382.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020947.mm.1 TC0900002495.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020947.mm.1 NM_017382.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020947.mm.1 NM_017382.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020950.mm.1 TC0900002495.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020950.mm.1 NM_017382.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020950.mm.1 NM_017382.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020945.mm.1 TC0900002495.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020945.mm.1 NM_017382.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020945.mm.1 NM_017382.5 Refseq
          Mouse Expression Set 430 1449256_a_at - -
          Mouse Expression Set 430 1449256_PM_a_at - -
          Mouse Expression Set 430 1434549_at - -
          Mouse Expression Set 430 1434549_PM_at - -
          Murine Genome U74v2 Set 96238_at - -
          Murine Genome U74v2 Set 96238_PM_at - -
          Murine Genome U74v2 Set 92854_at - -
          Murine Genome U74v2 Set 92854_PM_at - -
          Murine Genome U74v2 Set 167286_at - -
          Murine Genome U74v2 Set 167286_PM_at - -
          Mouse Genome 430A 2.0 Array 1449256_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1449256_a_at - -
          Mouse Genome 430A 2.0 Array 1434549_PM_at - -
          Mouse Genome 430A 2.0 Array 1434549_at - -
          Affymetrix HT Mouse Genome 430A Array 1449256_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449256_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1434549_at - -
          Affymetrix HT Mouse Genome 430A Array 1434549_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 359489 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100050402 - -
          Illumina mouse-6 v1.0 expression beadchip 100050402 - -
          Illumina mouse-6 v1.0 expression beadchip 5360079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230413 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258343 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230413 - -
          mouse-6 v1.1 (Illumina) scl35699.5_140-S - -
          mouse-6 v1.1 (Illumina) scl0053869.2_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 100050402 - -
          Illumina Sentrix mouse-6.v1 bead array 5360079 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0053869.2_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100050402 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360079 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230413 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230413 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0053869.2_7-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0