Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01171489_g1
          See other BNIP2 GE Assays ›
          Gene Symbol:
          Bnip2
          Gene Name:
          BCL2/adenovirus E1B interacting protein 2
          Gene Aliases:
          5730523P12Rik, BNIP2beta, Nip21
          Chromosome Location:
          Chr.9: 69989425 - 70011659 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01171489_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bnip2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008238.3 NP_001008239.1 1 - 2 223 103
          NM_016787.4 NP_058067.2 1 - 2 223 103
          GenBank mRNA AK014659.1 - 1 - 2 175 103
          AK054288.1 - 1 - 2 189 103
          AK159745.1 - 1 - 2 186 103
          AK161116.1 - 1 - 2 176 103
          AK169609.1 - 1 - 2 208 103
          BC012670.1 - 1 - 2 182 4037627 103

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK014659.1, AK054288.1, AK169609.1, BC012670.1, NM_001008238.3, NM_016787.4 transcripts.
          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bnip2

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst development
          apoptotic process
          centrosome cycle
          positive regulation of MAPK cascade
          positive regulation of neuron differentiation
          positive regulation of small GTPase mediated signal transduction
          striated muscle cell differentiation
          identical protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900007101.mm.1 TC0900000922.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007101.mm.1 NM_001008238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007101.mm.1 NM_016787.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007105.mm.1 TC0900000922.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007105.mm.1 NM_001008238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007105.mm.1 NM_016787.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007096.mm.1 TC0900000922.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007096.mm.1 NM_001008238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007096.mm.1 NM_016787.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007102.mm.1 TC0900000922.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007102.mm.1 NM_001008238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007102.mm.1 NM_016787.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007103.mm.1 TC0900000922.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007103.mm.1 NM_001008238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007103.mm.1 NM_016787.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007104.mm.1 TC0900000922.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007104.mm.1 NM_001008238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007104.mm.1 NM_016787.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007106.mm.1 TC0900000922.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007106.mm.1 NM_001008238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007106.mm.1 NM_016787.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 786699 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0