Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01172562_m1
          See other 4930563M21RIK GE Assays ›
          Gene Symbol:
          4930563M21Rik
          Gene Name:
          RIKEN cDNA 4930563M21 gene
          Gene Aliases:
          Rfpl3s
          Chromosome Location:
          Chr.9: 55962587 - 56026518 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01172562_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930563M21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183111.2 NP_898934.2 7 - 8 496 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002345.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 4930563M21Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137372 AK016210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137372 NM_183111.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2182557 AK016210.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30822 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 479773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940092 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415649 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 53670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138155 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174013 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169311 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2182557 AK016210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137372 AK016210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137372 NM_183111.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26533630 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06798066 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169311 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174013 - -
          Whole Mouse Genome G4122A A_52_P114802 AK016210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P114802 NM_183111.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114774 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174013 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169311 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08050595 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08050585 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08050588 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08050582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08050591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26533592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08050594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08011301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08007146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08075096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08075092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08075091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08007135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07503326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08007142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26533636 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10593835 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593836 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593839 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593840 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593838 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593841 - -
          Affymetrix Mouse Gene 1.1 ST Array 10593834 - -
          Affymetrix Mouse Clariom S Array PSR0900019687.mm.1 TC0900002345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900019687.mm.1 NM_183111.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900019687.mm.1 NM_183111.2 Refseq
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1780053 - -
          Illumina mouse-6 v1.0 expression beadchip 1780053 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780053 - -
          mouse-6 v1.1 (Illumina) scl35814.8.1_83-S - -
          Illumina Sentrix mouse-6.v1 bead array 1780053 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35814.8.1_83-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780053 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2960493 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228566 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2960493 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0