Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01172873_g1
          See other MFSD12 GE Assays ›
          Gene Symbol:
          Mfsd12
          Gene Name:
          major facilitator superfamily domain containing 12
          Gene Aliases:
          C19orf28, F630110N24Rik, Wdt1
          Chromosome Location:
          Chr.10: 81357469 - 81366225 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01172873_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mfsd12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028657.3 NP_082933.2 8 - 9 1303 116
          GenBank mRNA AK149678.1 - 8 - 9 1309 116
          AK154386.1 - 8 - 9 1305 116
          AK154902.1 - 8 - 9 1383 116
          AK170185.1 - 8 - 9 1322 116
          AK171084.1 - 8 - 9 1305 116
          BC027706.1 - 2 - 3 251 116
          BC038699.1 - 2 - 3 251 116
          BC118620.1 - 8 - 9 1303 40107417 116
          BC119790.1 - 8 - 9 1303 40107416 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000895.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289092 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289089 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289088 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289145 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289090 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289091 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289093 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006747 BC118620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006747 BC119790.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006747 AK171084.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006747 BC027706.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006747 BC038699.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006754 AK171084.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006754 AK154902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006754 AK149678.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233826 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573202 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114936 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105858 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174454 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006747 BC118620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006747 BC119790.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006747 AK171084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006747 BC027706.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006747 BC038699.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289165 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174454 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07125372 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105858 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174454 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105858 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289092 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289093 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289090 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289088 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289089 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289145 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08387853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08693860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08693879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08954089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08387857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08640401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08373538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08327489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08954093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08693874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08640404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289135 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10365112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365110 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365106 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365107 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365109 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365111 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365105 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365108 - -
          Affymetrix HT MG-430 PM Array Plate 1434263_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10365104 - -
          Mouse Genome 430 2.0 Array 1434263_at - -
          Mouse Genome 430 2.0 Array 1434263_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73822_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73822_at - -
          Affymetrix Mouse Clariom S Array PSR1000007068.mm.1 TC1000000895.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007068.mm.1 NM_028657.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007068.mm.1 NM_028657.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007067.mm.1 TC1000000895.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007067.mm.1 NM_028657.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007067.mm.1 NM_028657.3 Refseq
          Mouse Expression Set 430 1434263_at - -
          Mouse Expression Set 430 1434263_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434263_at - -
          Affymetrix HT Mouse Genome 430B Array 1434263_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242469 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242469 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0