Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01172875_g1
          See other FZR1 GE Assays ›
          Gene Symbol:
          Fzr1
          Gene Name:
          fizzy/cell division cycle 20 related 1 (Drosophila)
          Gene Aliases:
          AW108046, Cdh1, FZR, FZR2, Fyr, HCDH, HCDH1
          Chromosome Location:
          Chr.10: 81366879 - 81378370 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01172875_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fzr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019757.1 NP_062731.1 11 - 12 1385 67
          GenBank mRNA AF083809.1 - 11 - 12 1385 67
          AK051993.1 - 11 - 12 1308 67
          AK081476.1 - 11 - 12 1441 67
          AK083686.1 - 1455 67
          AK154822.1 - 11 - 12 1469 67
          AK155370.1 - 11 - 12 1467 67
          AK163951.1 - 11 - 12 1444 67
          AK172679.1 - 8 - 9 2405 67
          AK178744.1 - 2 - 3 307 67
          AK209977.1 - 2 - 3 307 67
          AK212079.1 - 1 - 2 106 67
          AK214012.1 - 1 - 2 107 67
          BC006616.1 - 11 - 12 1466 3967009 67
          FJ786633.2 - 6 - 7 619 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002540.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fzr1

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          DNA repair
          cellular response to DNA damage stimulus
          cell cycle
          positive regulation of cell population proliferation
          anaphase-promoting complex-dependent catabolic process
          regulation of meiotic nuclear division
          positive regulation of protein catabolic process
          cell division
          lens fiber cell differentiation
          negative regulation of cell aging
          positive regulation of ubiquitin protein ligase activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000019066.mm.1 TC1000002540.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000019066.mm.1 NM_019757.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000019066.mm.1 NM_019757.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000019064.mm.1 TC1000002540.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000019064.mm.1 NM_019757.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000019064.mm.1 NM_019757.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0