Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01172900_g1
          See other NMRK2 GE Assays ›
          Gene Symbol:
          Nmrk2
          Gene Name:
          nicotinamide riboside kinase 2
          Gene Aliases:
          2310015C21Rik, Itgb1bp3, Mibp
          Chromosome Location:
          Chr.10: 81198170 - 81202212 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01172900_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nmrk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027120.2 NP_081396.1 6 - 7 564 126
          GenBank mRNA AK009352.1 - 6 - 7 580 126
          AK137483.1 - 6 - 7 566 126
          BC119244.1 - 6 - 7 526 8733993 126
          BC119246.1 - 6 - 7 526 8733995 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002530.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphorylation
          pyridine nucleotide biosynthetic process
          negative regulation of myoblast differentiation
          nucleotide binding
          protein binding
          ATP binding
          kinase activity
          transferase activity
          metal ion binding
          ribosylnicotinamide kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369762 AK009352.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369762 AK137483.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369762 BC119244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369762 BC119246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369762 NM_027120.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369762 AK009352.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369762 AK137483.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369762 BC119244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369762 BC119246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369762 NM_027120.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 708522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184179 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29747 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135932 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369762 AK009352.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369762 AK137483.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369762 BC119244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369762 BC119246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369762 NM_027120.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369762 AK009352.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369762 AK137483.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369762 BC119244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369762 BC119246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369762 NM_027120.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369762 AK009352.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369762 AK137483.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369762 BC119244.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369762 BC119246.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369762 NM_027120.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369762 AK009352.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369762 AK137483.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369762 BC119244.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369762 BC119246.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369762 NM_027120.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369762 AK009352.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369762 AK137483.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369762 BC119244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369762 BC119246.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369762 NM_027120.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369762 AK009352.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369762 AK137483.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369762 BC119244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369762 BC119246.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369762 NM_027120.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08752517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08470659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08633713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08633704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08325291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08943700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08943703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08698313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08312027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08724533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08793632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08633708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288055 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10371084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371091 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371090 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371086 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371087 - -
          Affymetrix HT MG-430 PM Array Plate 1453898_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10371082 - -
          Mouse Genome 430 2.0 Array 1453898_at - -
          Mouse Genome 430 2.0 Array 1453898_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69564_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69564_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000018921.mm.1 TC1000002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018921.mm.1 NM_027120.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018921.mm.1 NM_027120.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018920.mm.1 TC1000002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018920.mm.1 NM_027120.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018920.mm.1 NM_027120.2 Refseq
          Mouse Expression Set 430 1453898_at - -
          Mouse Expression Set 430 1453898_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453898_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453898_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 315822 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2724880 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2727022 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2727022 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2724880 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244715 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0