Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175595_m1
          See other TCF3 GE Assays ›
          Gene Symbol:
          Tcf3
          Gene Name:
          transcription factor 3
          Gene Aliases:
          A1, AA408400, ALF2, AW209082, E12, E12/E47, E2A, E47, ME2, Pan1, Pan2, TCF-3, Tcfe2a, VDIR, bHLHb21
          Chromosome Location:
          Chr.10: 80409165 - 80433653 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175595_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcf3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164147.1 NP_001157619.1 5 - 6 698 70
          NM_001164148.1 NP_001157620.1 5 - 6 698 70
          NM_001164149.1 NP_001157621.1 5 - 6 698 70
          NM_001164150.1 NP_001157622.1 5 - 6 698 70
          NM_001164151.1 NP_001157623.1 5 - 6 698 70
          NM_001164152.1 NP_001157624.1 5 - 6 698 70
          NM_001164153.1 NP_001157625.1 5 - 6 698 70
          NM_011548.4 NP_035678.3 5 - 6 698 70
          GenBank mRNA AF352579.1 - 5 - 6 393 70
          AK017617.1 - 5 - 6 666 70
          AK034777.1 - 5 - 6 681 70
          AK038738.1 - 5 - 6 685 70
          AK146786.1 - 5 - 6 70
          AK147735.1 - 4 - 5 354 70
          AK156265.1 - 5 - 6 681 70
          AK161009.1 - 5 - 6 683 70
          AK170674.1 - 5 - 6 692 70
          BC006860.1 - 5 - 6 652 2631291 70
          BC018260.1 - 4 - 5 323 4187386 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002501.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcf3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          natural killer cell differentiation
          B cell lineage commitment
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          gastrulation
          nervous system development
          response to xenobiotic stimulus
          positive regulation of gene expression
          lymphocyte differentiation
          cell differentiation
          positive regulation of B cell proliferation
          response to lipopolysaccharide
          T cell differentiation in thymus
          immunoglobulin V(D)J recombination
          histone H3 acetylation
          histone H4 acetylation
          positive regulation of neuron differentiation
          positive regulation of cell cycle
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          cell development
          Peyer's patch development
          protein stabilization
          positive regulation of DNA-binding transcription factor activity
          regulation of G1/S transition of mitotic cell cycle
          transcription cis-regulatory region binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA binding
          chromatin binding
          DNA-binding transcription factor activity
          transcription coactivator activity
          protein binding
          transcription factor binding
          PDZ domain binding
          mitogen-activated protein kinase kinase kinase binding
          protein homodimerization activity
          bHLH transcription factor binding
          sequence-specific DNA binding
          protein heterodimerization activity
          protein dimerization activity
          vitamin D response element binding
          E-box binding
          DNA-binding transcription factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 TC1000002501.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164147.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164147.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164148.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164148.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164149.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164149.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164150.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164150.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164151.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164151.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164153.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_001164153.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_011548.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018543.mm.1 NM_011548.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0