Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175698_m1
          See other E130310I04RIK GE Assays ›
          Gene Symbol:
          E130310I04Rik
          Gene Name:
          RIKEN cDNA E130310I04 gene
          Gene Aliases:
          A930019C19Rik
          Chromosome Location:
          Chr.16: 34943175 - 34958994 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175698_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E130310I04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045722.1 1 - 2 101 61
          NR_045723.1 1 - 2 231 61
          GenBank mRNA AK044449.1 - 1 - 2 116 61
          AK053822.1 - 1 - 2 102 61
          CK618313.1 - 1 - 2 231 61
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB642401.1 - 1 - 2 109 Chr. 16 - 34954866 - 34958872 [-] on Build GRCm38 61
          GenBankmRNA BF463941.1 - 1 - 2 186 Chr. 16 - 34955068 - 34958892 [-] on Build GRCm38 61
          GenBankmRNA BB665513.1 - 1 - 2 98 Chr. 16 - 34954881 - 34958861 [-] on Build GRCm38 61

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream myosin,
          light polypeptide kinase
          Mylk 9530072E15Rik,
          A930019C19Rik,
          AW489456,
          KRP,
          MLCK108,
          MLCK210,
          Mlck,
          nmMlck,
          smMLCK
          Anti-sense Chr. 16 - 34745199 - 35002436 on NCBI Build GRCm38
          Downstream 3-hydroxyacyl-CoA dehydratase 2 Hacd2 6330408J20Rik,
          AI255777,
          AI481689,
          Hcad2,
          Ptplb
          63.5kb Anti-sense Chr. 16 - 35022421 - 35109190 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001540.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111679 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600012667.mm.1 TC1600001540.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012667.mm.1 NR_045722.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012667.mm.1 NR_045722.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012666.mm.1 TC1600001540.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012666.mm.1 NR_045723.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012666.mm.1 NR_045723.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012668.mm.1 TC1600001540.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012668.mm.1 NR_045722.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012668.mm.1 NR_045722.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012668.mm.1 NR_045723.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012668.mm.1 NR_045723.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012665.mm.1 TC1600001540.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012665.mm.1 NR_045723.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012665.mm.1 NR_045723.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012670.mm.1 TC1600001540.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012670.mm.1 NR_045723.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012670.mm.1 NR_045723.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012669.mm.1 TC1600001540.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012669.mm.1 NR_045722.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012669.mm.1 NR_045722.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012669.mm.1 NR_045723.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012669.mm.1 NR_045723.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 431704 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5756d5bff6-86w5v:80/100.66.130.62:80.
          git-commit: 27702359083d4c7b0962b032f2611a59cac079b5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0