Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287217.1 | NP_001274146.1 | 1 - 2 | 242 | 66 | |
NM_008761.4 | NP_032787.1 | 1 - 2 | 242 | 66 | ||
GenBank mRNA | AI121044.1 | - | 1 - 2 | 242 | 66 | |
CA528582.1 | - | 1 - 2 | 233 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() |