Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01176407_m1
          See other TBRG1 GE Assays ›
          Gene Symbol:
          Tbrg1
          Gene Name:
          transforming growth factor beta regulated gene 1
          Gene Aliases:
          AA408552, AA409675, Niam, Tb-5, mFLJ00213
          Chromosome Location:
          Chr.9: 37649182 - 37657312 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01176407_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tbrg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025289.3 NP_079565.2 1 - 2 314 67
          GenBank mRNA AK010925.1 - 1 - 2 265 67
          AK011268.1 - 1 - 2 238 67
          AK011614.1 - 1 - 2 260 67
          AK131158.1 - 1 - 2 243 67
          AK131638.1 - 1 - 2 160 67
          AK150549.1 - 1 - 2 241 67
          AK151075.1 - 1 - 2 192 67
          AK151209.1 - 1 - 2 208 67
          AK151457.1 - 1 - 2 226 67
          AK166547.1 - 1 - 2 236 67
          BC021331.1 - 1 - 2 325 5003215 67
          BC037145.1 - 1 - 2 233 67
          BC040813.1 - 1 - 2 227 4223194 67
          BC043022.1 - 1 - 2 229 5688749 67
          BC054057.1 - 1 - 2 224 67
          BC065795.1 - 1 - 2 227 1529903 67
          DQ144541.1 - 1 - 2 146 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002054.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tbrg1

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          cell cycle
          negative regulation of cell population proliferation
          protein stabilization
          protein localization to nucleoplasm
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900017148.mm.1 TC0900002054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900017148.mm.1 NM_025289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017148.mm.1 NM_025289.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017151.mm.1 TC0900002054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900017151.mm.1 NM_025289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017151.mm.1 NM_025289.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017154.mm.1 TC0900002054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900017154.mm.1 NM_025289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017154.mm.1 NM_025289.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017149.mm.1 TC0900002054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900017149.mm.1 NM_025289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017149.mm.1 NM_025289.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 866620 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0