Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01176518_g1
          See other CD22 GE Assays ›
          Gene Symbol:
          Cd22
          Gene Name:
          CD22 antigen
          Gene Aliases:
          A530093D23, Lyb-8, Lyb8
          Chromosome Location:
          Chr.7: 30865404 - 30880342 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01176518_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001043317.2 NP_001036782.1 13 - 14 2640 100
          NM_009845.3 NP_033975.3 11 - 12 2515 100
          NR_102722.1 13 - 14 2712 100
          NR_102723.1 13 - 14 2673 100
          GenBank mRNA AF102134.1 - 10 - 11 2284 100
          AF115400.1 - 10 - 11 2284 100
          AF115401.1 - 10 - 11 2284 100
          AK041230.1 - 13 - 14 2647 100
          AK143835.1 - 13 - 14 2578 100
          AK156735.1 - 11 - 12 2514 100
          AK171589.1 - 10 - 11 2321 100
          AK172301.1 - 11 - 12 2471 100
          BC046957.1 - 13 - 14 2585 100
          BC150807.1 - 12 - 13 2599 9087718 100
          L02844.1 - 12 - 13 2786 100
          L16928.1 - 12 - 13 2763 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002779.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd22

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          cell surface receptor signaling pathway
          protein binding
          coreceptor activity
          carbohydrate binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700024807.mm.1 TC0700002779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024807.mm.1 NM_001043317.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024807.mm.1 NM_001043317.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024807.mm.1 NM_009845.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024807.mm.1 NM_009845.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024807.mm.1 NR_102722.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024807.mm.1 NR_102723.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024806.mm.1 TC0700002779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024806.mm.1 NM_001043317.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024806.mm.1 NM_001043317.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024806.mm.1 NM_009845.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024806.mm.1 NM_009845.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024806.mm.1 NR_102722.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024806.mm.1 NR_102723.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0