Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104616.1 | NP_001098086.1 | 8 - 9 | 941 | 120 | |
NM_001104617.1 | NP_001098087.1 | 8 - 9 | 1347 | 120 | ||
NM_009041.3 | NP_033067.2 | 8 - 9 | 1347 | 120 | ||
GenBank mRNA | AK016711.1 | - | 8 - 9 | 1017 | 120 | |
AK088292.1 | - | 8 - 9 | 1054 | 120 | ||
AK156370.1 | - | 8 - 9 | 1033 | 120 | ||
AK162204.1 | - | 8 - 9 | 919 | 120 | ||
AK168449.1 | - | 8 - 9 | 1018 | 120 | ||
AK206105.1 | - | 2 - 3 | 120 | |||
BC002041.1 | - | 8 - 9 | 992 | 120 | ||
BC026786.1 | - | 8 - 9 | 1020 | 120 | ||
BC034438.1 | - | 8 - 9 | 1173 | 120 | ||
BC053417.1 | - | 8 - 9 | 1021 | 6707873 | 120 | |
X60672.1 | - | 8 - 9 | 960 | 120 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |