Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01178151_m1
          See other ADAT3 GE Assays ›
          Gene Symbol:
          Adat3
          Gene Name:
          adenosine deaminase, tRNA-specific 3
          Gene Aliases:
          A430024H01Rik
          Chromosome Location:
          Chr.10: 80602880 - 80607654 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01178151_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adat3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100606.1 NP_001094076.1 1 - 2 71 77
          GenBank mRNA BY036102.1 - 1 - 2 71 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000864.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001100606.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA processing
          biological_process
          molecular_function
          catalytic activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2182392 NM_001100606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2182392 BX632866.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687486 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513138 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 158064 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2182392 NM_001100606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2182392 BX632866.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27284216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27284223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27284229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27284237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27284226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27284227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27284231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27284225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27284235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27284228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08429300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27284234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27284233 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364816 - -
          Affymetrix HT MG-430 PM Array Plate 1430327_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10364814 - -
          Mouse Genome 430 2.0 Array 1430327_PM_at - -
          Mouse Genome 430 2.0 Array 1430327_at - -
          Affymetrix Mouse Clariom S Array PSR1000006561.mm.1 TC1000000864.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000006561.mm.1 NM_001100606.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006561.mm.1 NM_001100606.1 Refseq
          Mouse Expression Set 430 1450393_PM_a_at - -
          Mouse Expression Set 430 1421842_a_at - -
          Mouse Expression Set 430 1450393_a_at - -
          Mouse Expression Set 430 1421842_PM_a_at - -
          Mouse Expression Set 430 1430327_PM_at - -
          Mouse Expression Set 430 1430327_at - -
          Murine Genome U74v2 Set 171292_PM_at - -
          Murine Genome U74v2 Set 171292_at - -
          Murine Genome U74v2 Set 170279_PM_r_at - -
          Murine Genome U74v2 Set 170279_r_at - -
          Mouse Genome 430A 2.0 Array 1421842_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1450393_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1450393_a_at - -
          Mouse Genome 430A 2.0 Array 1421842_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1430327_at - -
          Affymetrix HT Mouse Genome 430B Array 1430327_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103060377 - -
          Illumina mouse-6 v1.0 expression beadchip 103060377 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248910 - -
          Illumina Sentrix mouse-6.v1 bead array 103060377 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103060377 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2550740 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0