Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01178945_g1
          See other RDH8 GE Assays ›
          Gene Symbol:
          Rdh8
          Gene Name:
          retinol dehydrogenase 8
          Gene Aliases:
          Gm182, prRDH
          Chromosome Location:
          Chr.9: 20818504 - 20826163 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01178945_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rdh8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001030290.1 NP_001025461.1 5 - 6 722 84

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rdh8

          Gene Ontology Categories:

          Function(s) Process(es)

          visual perception
          retinol metabolic process
          NAD-retinol dehydrogenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1970570 NM_001030290.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536607 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40150 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108066 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1970570 NM_001030290.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26344063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26344066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26344067 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06717035 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108066 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108066 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07682238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26344054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08129062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07682239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07665006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07473430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07456183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07490423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08122538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08172091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07665005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26344076 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583479 - -
          Affymetrix Mouse Gene 1.1 ST Array 10583478 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 632118 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2533386 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2860731 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2860731 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0