Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179050_g1
          See other RINL GE Assays ›
          Gene Symbol:
          Rinl
          Gene Name:
          Ras and Rab interactor-like
          Gene Aliases:
          5830482F20Rik, 9930116N10Rik
          Chromosome Location:
          Chr.7: 28788861 - 28798965 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179050_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rinl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177158.5 NP_796132.2 11 - 12 1717 77
          GenBank mRNA AK082828.1 - 4 - 5 953 77
          AK162469.1 - 4 - 5 622 77
          AK168472.1 - 4 - 5 880 77
          BC044761.1 - 11 - 12 1753 77
          BC044878.1 - 2 - 3 434 77
          BC046530.1 - 11 - 12 1753 5329601 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000512.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rinl

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          endocytosis
          protein transport
          positive regulation of GTPase activity
          guanyl-nucleotide exchange factor activity
          GTPase activator activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487918 BC046530.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487918 BF147913.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487918 AK168472.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487918 BC044761.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487918 BC044878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487918 BC046530.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487918 BF147913.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487918 AK168472.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487918 BC044761.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487918 BC044878.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256096 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707092 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431758 AF539600.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431758 AY211081.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431758 NM_177634.1 refseq_mrna_mouse_NM
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163010 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487918 BC046530.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487918 BF147913.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487918 AK168472.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487918 BC044761.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487918 BC044878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487918 BC046530.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487918 BF147913.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487918 AK168472.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487918 BC044761.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487918 BC044878.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023602 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163010 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01490601 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06150398 - -
          Whole Mouse Genome G4122A A_51_P294566 AK020687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P25548 BC046530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P25548 AK168472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P25548 BC044761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P25548 BC044878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P25548 AK030993.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487918 BC046530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487918 BF147913.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487918 AK168472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487918 BC044761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487918 BC044878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487918 BC046530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487918 BF147913.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487918 AK168472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487918 BC044761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487918 BC044878.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134095 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121883 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116738 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163010 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01490601 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06111935 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06327315 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06327324 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06327310 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06117607 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06111940 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06111950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06327319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06574388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05982321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06579905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06111950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06114395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06574387 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551592 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551590 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551591 - -
          Affymetrix HT MG-430 PM Array Plate 1455576_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1431100_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1444197_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551587 - -
          Mouse Genome 430 2.0 Array 1455576_at - -
          Mouse Genome 430 2.0 Array 1444197_at - -
          Mouse Genome 430 2.0 Array 1444197_PM_at - -
          Mouse Genome 430 2.0 Array 1455576_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320435_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320435_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700004215.mm.1 TC0700000512.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004215.mm.1 NM_177158.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004215.mm.1 NM_177158.5 Refseq
          Mouse Expression Set 430 1455576_PM_at - -
          Mouse Expression Set 430 1444197_at - -
          Mouse Expression Set 430 1455576_at - -
          Mouse Expression Set 430 1444197_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431100_at - -
          Affymetrix HT Mouse Genome 430B Array 1444197_at - -
          Affymetrix HT Mouse Genome 430B Array 1431100_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455576_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444197_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455576_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3440341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2662317 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2662317 - -
          mouse-6 v1.1 (Illumina) scl0320435.14_117-S - -
          Illumina Sentrix mouse-6.v1 bead array 3440341 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0320435.14_117-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2662316 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2662316 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0