Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179677_m1
          See other SERINC1 GE Assays ›
          Gene Symbol:
          Serinc1
          Gene Name:
          serine incorporator 1
          Gene Aliases:
          1500011D18Rik, AI315070, AIGP-2, TMS-2, Tde1l, Tde2, Tms2, mKIAA1253
          Chromosome Location:
          Chr.10: 57515774 - 57532529 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179677_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serinc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019760.3 NP_062734.1 1 - 2 169 121
          GenBank mRNA AB029500.1 - 1 - 2 113 121
          AK002847.1 - 1 - 2 118 121
          AK050514.1 - 1 - 2 110 121
          AK133668.1 - 1 - 2 110 121
          AK133975.1 - 1 - 2 110 121
          AK141574.1 - 1 - 2 118 121
          AK159436.1 - 1 - 2 136 121
          AK160498.1 - 1 - 2 117 121
          AK161707.1 - 1 - 2 111 121
          AK167817.1 - 1 - 2 118 121
          BC006940.1 - 1 - 2 144 121
          CJ142862.1 - 1 - 2 171 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002195.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AB029500.1, AK002847.1, AK133668.1, AK133975.1, AK141574.1, AK159436.1, NM_019760.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serinc1

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          phosphatidylserine metabolic process
          sphingolipid metabolic process
          phospholipid biosynthetic process
          L-serine transport
          positive regulation of transferase activity
          cation transmembrane transport
          L-alpha-amino acid transmembrane transport
          L-serine transmembrane transporter activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000015404.mm.1 TC1000002195.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015404.mm.1 NM_019760.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015404.mm.1 NM_019760.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015400.mm.1 TC1000002195.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015400.mm.1 NM_019760.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015400.mm.1 NM_019760.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015405.mm.1 TC1000002195.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015405.mm.1 NM_019760.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015405.mm.1 NM_019760.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015399.mm.1 TC1000002195.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015399.mm.1 NM_019760.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015399.mm.1 NM_019760.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 906008 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0