Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmym1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026670.4 | NP_080946.3 | 1 - 2 | 135 | 197 | |
GenBank mRNA | AK016783.1 | - | 1 - 2 | 139 | 197 | |
AK035618.1 | - | 1 - 2 | 135 | 197 | ||
AK044538.1 | - | 1 - 2 | 145 | 197 | ||
AK049568.1 | - | 1 - 2 | 125 | 197 | ||
AK138687.1 | - | 1 - 2 | 97 | 197 | ||
AK166769.1 | - | 1 - 2 | 118 | 197 | ||
AK167508.1 | - | 1 - 2 | 141 | 197 | ||
BC027750.1 | - | 1 - 2 | 162 | 197 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |