Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179916_g1
          See other OGDH GE Assays ›
          Gene Symbol:
          Ogdh
          Gene Name:
          oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide)
          Gene Aliases:
          2210403E04Rik, 2210412K19Rik, AA409584, d1401, mKIAA4192
          Chromosome Location:
          Chr.11: 6291593 - 6359094 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179916_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ogdh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252282.1 NP_001239211.1 12 - 13 1682 76
          NM_001252283.1 NP_001239212.1 12 - 13 1670 76
          NM_001252287.1 NP_001239216.1 11 - 12 1713 76
          NM_001252288.1 NP_001239217.1 11 - 12 1685 76
          NM_010956.4 NP_035086.2 11 - 12 1637 76
          GenBank mRNA AK147289.1 - 11 - 12 1638 76
          AK150009.1 - 11 - 12 1603 76
          AK169286.1 - 11 - 12 1629 76
          AK191182.1 - 1 - 2 149 76
          AK193717.1 - 149 76
          AK195375.1 - 1 - 2 149 76
          AK216535.1 - 1 - 2 149 76
          AK220536.1 - 12 - 13 1644 76
          BC013670.1 - 11 - 12 1571 4188988 76
          BC049104.1 - 11 - 12 1638 6417392 76
          BC057354.1 - 12 - 13 1598 6827509 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000093.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ogdh

          Gene Ontology Categories:

          Function(s) Process(es)

          glycolytic process
          tricarboxylic acid cycle
          2-oxoglutarate metabolic process
          succinyl-CoA metabolic process
          NADH metabolic process
          metabolic process
          oxoglutarate dehydrogenase (succinyl-transferring) activity
          oxidoreductase activity
          oxidoreductase activity, acting on the aldehyde or oxo group of donors, disulfide as acceptor
          thiamine pyrophosphate binding
          heat shock protein binding
          oxoglutarate dehydrogenase (NAD+) activity
          metal ion binding
          chaperone binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100000982.mm.1 TC1100000093.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000982.mm.1 NM_001252282.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000982.mm.1 NM_001252282.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000982.mm.1 NM_001252283.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000982.mm.1 NM_001252283.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000982.mm.1 NM_001252287.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000982.mm.1 NM_001252287.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000982.mm.1 NM_001252288.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000982.mm.1 NM_001252288.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000982.mm.1 NM_010956.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000982.mm.1 NM_010956.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000983.mm.1 TC1100000093.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000983.mm.1 NM_001252282.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000983.mm.1 NM_001252282.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000983.mm.1 NM_001252283.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000983.mm.1 NM_001252283.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000983.mm.1 NM_001252287.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000983.mm.1 NM_001252287.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000983.mm.1 NM_001252288.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000983.mm.1 NM_001252288.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000983.mm.1 NM_010956.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000983.mm.1 NM_010956.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline