Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179996_m1
          See other SLC18B1 GE Assays ›
          Gene Symbol:
          Slc18b1
          Gene Name:
          solute carrier family 18, subfamily B, member 1
          Gene Aliases:
          1110021L09Rik
          Chromosome Location:
          Chr.10: 23796654 - 23827969 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179996_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc18b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183116.2 NP_898939.2 11 - 12 1314 107
          GenBank mRNA BC068251.1 - 6 - 7 579 107
          BM949736.1 - 5 - 6 498 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000191.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          transmembrane transport
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955289 AK003902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955289 NM_183116.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 280690 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268333 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 821854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487610 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924673 AK009371.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924673 AK010055.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924673 AK010107.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924673 BC031537.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 87822 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919322 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507128 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 326383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15644 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42600 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149981 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955289 AK003902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955289 NM_183116.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149981 - -
          Whole Mouse Genome G4122A A_51_P369381 AK003902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369381 NM_183116.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27005360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08652128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08836002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08515172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08825057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08853106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08842883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08698654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08882067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08924521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08611848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08925425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08652131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08825058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08368679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08276245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08769078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08886246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08609697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08620241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27005348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08610743 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10362127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362122 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362128 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362119 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362121 - -
          Affymetrix HT MG-430 PM Array Plate 1428795_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10362115 - -
          Mouse Genome 430 2.0 Array 1428795_at - -
          Mouse Genome 430 2.0 Array 1428795_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76306_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76306_at - -
          Affymetrix Mouse Clariom S Array PSR1000001415.mm.1 TC1000000191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000001415.mm.1 NM_183116.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001415.mm.1 NM_183116.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001414.mm.1 TC1000000191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000001414.mm.1 NM_183116.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001414.mm.1 NM_183116.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001416.mm.1 TC1000000191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000001416.mm.1 NM_183116.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001416.mm.1 NM_183116.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001413.mm.1 TC1000000191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000001413.mm.1 NM_183116.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001413.mm.1 NM_183116.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001412.mm.1 TC1000000191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000001412.mm.1 NM_183116.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001412.mm.1 NM_183116.2 Refseq
          Mouse Expression Set 430 1428795_at - -
          Mouse Expression Set 430 1428795_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428795_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428795_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 326153 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1340594 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2738655 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2738655 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0