Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01180529_m1
          See other RNF217 GE Assays ›
          Gene Symbol:
          Rnf217
          Gene Name:
          ring finger protein 217
          Gene Aliases:
          AU016819, Ibrdc1
          Chromosome Location:
          Chr.10: 31501887 - 31609725 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01180529_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf217
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146349.1 NP_001139821.1 5 - 6 2010 100
          GenBank mRNA CN459606.1 - 4 - 5 552 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001962.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P3845 BG070699.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P3845 NM_001146349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P3845 AI853829.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P3845 BG070699.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P3845 NM_001146349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P3845 AI853829.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128577 CN459606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128577 NM_001146349.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256709 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613839 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 341006 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789353 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34227 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582956 AK016480.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582956 AK028498.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582956 AK038320.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582956 AK122560.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582956 AK141804.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582956 AK163345.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582956 BC043127.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582956 BC055330.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582956 BC057374.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582956 NM_027642.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838130 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 179230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 927055 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 588511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127923 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173369 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 243148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172047 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 857252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714205 BC127964.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714205 NM_207150.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71387 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 857108 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630212 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 95094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 160183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940370 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395406 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945378 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59767 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9740 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43713 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8717 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50975 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170282 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136827 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P3845 BG070699.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P3845 NM_001146349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P3845 AI853829.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P3845 BG070699.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P3845 NM_001146349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P3845 AI853829.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128577 CN459606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128577 NM_001146349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450854 CN459606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450854 NM_001146349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450854 AK219068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450854 AK219870.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P3845 BG070699.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P3845 NM_001146349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P3845 AI853829.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P3845 BG070699.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P3845 NM_001146349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P3845 AI853829.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08223694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08561843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08670477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08561847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08410924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08485142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08670478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27044913 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10368581 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368583 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368582 - -
          Affymetrix HT MG-430 PM Array Plate 1440179_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1436305_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10368577 - -
          Mouse Genome 430 2.0 Array 1436305_at - -
          Mouse Genome 430 2.0 Array 1440179_PM_x_at - -
          Mouse Genome 430 2.0 Array 1436305_PM_at - -
          Mouse Genome 430 2.0 Array 1440179_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 268291_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 268291_at - -
          Affymetrix Mouse Clariom S Array PSR1000014074.mm.1 TC1000001962.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000014074.mm.1 NM_001146349.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014074.mm.1 NM_001146349.1 Refseq
          Mouse Expression Set 430 1440179_PM_x_at - -
          Mouse Expression Set 430 1436305_PM_at - -
          Mouse Expression Set 430 1436305_at - -
          Mouse Expression Set 430 1440179_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1440179_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1440179_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1436305_at - -
          Affymetrix HT Mouse Genome 430B Array 1436305_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 873206 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0