Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01180776_g1
          See other SNHG3 GE Assays ›
          Gene Symbol:
          Snhg3
          Gene Name:
          small nucleolar RNA host gene 3
          Gene Aliases:
          Rnu17d, U17HG
          Chromosome Location:
          Chr.4: 132351933 - 132353686 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01180776_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snhg3
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003270.2
          2 - 3 119 117
          GenBank mRNA AJ006837.1 - LIT1313
          U17HG
          2 - 3 66 117
          AK076188.1 -
          2 - 3 67 117
          BC100513.1 -
          2 - 3 112 117
          CN834544.1 -
          2 - 3 122 117
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA EL959114.1 - 2 - 3 111 Chr. 4 - 132351934 - 132353651 [-] on Build GRCm38 117
          GenBankmRNA BY118837.1 - 2 - 3 103 Chr. 4 - 132352035 - 132353668 [-] on Build GRCm38 117
          GenBankmRNA CN841655.1 - 2 - 3 115 Chr. 4 - 132351934 - 132353679 [-] on Build GRCm38 117
          GenBankmRNA DV069876.1 - 135 Chr. - - [-] on Build 117
          GenBankmRNA DV069651.1 - 86 Chr. - - [-] on Build 117
          GenBankmRNA DV061498.1 - 164 Chr. - - [-] on Build 117
          GenBankmRNA CA460116.1 - 156 Chr. - - [-] on Build 117
          GenBankmRNA DV062701.1 - 187 Chr. - - [-] on Build 117
          GenBankmRNA AI787530.1 - 2 - 3 65 Chr. 4 - 132351934 - 132353632 [-] on Build GRCm38 117
          GenBankmRNA CJ216122.1 - 1 - 2 81 Chr. 4 - 132351934 - 132352750 [-] on Build GRCm38 117
          GenBankmRNA CA456802.1 - 133 Chr. - - [-] on Build 117
          GenBankmRNA BY144490.1 - 2 - 3 63 Chr. 4 - 132351966 - 132353630 [-] on Build GRCm38 117
          GenBankmRNA BY121084.1 - 2 - 3 102 Chr. 4 - 132352035 - 132353668 [-] on Build GRCm38 117
          GenBankmRNA BX639589.1 - 1 - 2 Chr. 4 - 132351934 - 132353658 [+] on Build GRCm38 117
          GenBankmRNA BB605517.1 - 2 - 3 69 Chr. 4 - 132352034 - 132353634 [-] on Build GRCm38 117
          GenBankmRNA DV072144.1 - 86 Chr. - - [-] on Build 117
          GenBankmRNA BE198161.1 - 2 - 3 93 Chr. 4 - 132352117 - 132353660 [-] on Build GRCm38 117
          GenBankmRNA CA492396.1 - 103 Chr. - - [-] on Build 117
          GenBankmRNA BY713659.1 - 2 - 3 61 Chr. 4 - 132351933 - 132353628 [-] on Build GRCm38 117
          GenBankmRNA AI788223.1 - 2 - 3 65 Chr. 4 - 132351934 - 132353632 [-] on Build GRCm38 117
          GenBankmRNA BY792896.1 - 2 - 3 97 Chr. 4 - 132348014 - 132353663 [-] on Build GRCm38 117
          GenBankmRNA BY579581.1 - 1 - 2 107 Chr. 4 - 132351934 - 132352776 [-] on Build GRCm38 117
          GenBankmRNA DV041333.1 - 161 Chr. - - [-] on Build 117
          GenBankmRNA EL964328.1 - 1 - 2 Chr. 4 - 132351934 - 132353658 [+] on Build GRCm38 117
          GenBankmRNA CA786577.1 - 115 Chr. - - [-] on Build 117

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA,
          H/ACA box 73b
          Snora73b U17B Sense Chr. 4 - 132352322 - 132352526 on NCBI Build GRCm38
          Downstream small nucleolar RNA,
          H/ACA box 73a
          Snora73a E1,
          E1-7,
          E1b,
          U17A
          77.0 Sense Chr. 4 - 132352775 - 132352979 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004223.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P661071 NR_003270.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P661071 AJ006837.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P661071 AK076188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P661071 BC100513.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P661071 CN834544.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P661071 NR_003270.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P661071 AJ006837.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P661071 AK076188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P661071 BC100513.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P661071 CN834544.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140421 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P661071 NR_003270.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P661071 AJ006837.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P661071 AK076188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P661071 BC100513.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P661071 CN834544.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P661071 NR_003270.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P661071 AJ006837.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P661071 AK076188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P661071 BC100513.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P661071 CN834544.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1433789_PM_at - -
          Mouse Genome 430 2.0 Array 1433789_PM_at - -
          Mouse Genome 430 2.0 Array 1433789_at - -
          Affymetrix Mouse Clariom S Array PSR0400030077.mm.1 TC0400004223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030077.mm.1 NR_003270.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030077.mm.1 NR_003270.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030076.mm.1 TC0400004223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030076.mm.1 NR_003270.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030076.mm.1 NR_003270.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030075.mm.1 TC0400004223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030075.mm.1 NR_003270.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030075.mm.1 NR_003270.2 Refseq
          Mouse Expression Set 430 1433789_at - -
          Mouse Expression Set 430 1433789_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433789_at - -
          Affymetrix HT Mouse Genome 430B Array 1433789_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 579357 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline