Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01181703_m1
          See other PISD-PS3 GE Assays ›
          Gene Symbol:
          Pisd-ps3
          Gene Name:
          phosphatidylserine decarboxylase, pseudogene 3
          Gene Aliases:
          4933439C20Rik
          Chromosome Location:
          Chr.Un|NT_187064: 52674 - 59689 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01181703_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pisd-ps3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK081907.1 - 5 - 6 2443 61
          AK168892.1 - 6 - 7 2201 61

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Mus musculus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122158 BC034366.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122158 BC048401.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122158 AK168892.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122158 BC003217.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122158 AK144226.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373703 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373704 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373706 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373708 - -
          Affymetrix HT MG-430 PM Array Plate 1439069_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1434975_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1453144_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453145_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429452_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373702 - -
          Mouse Genome 430 2.0 Array 1439069_PM_a_at - -
          Mouse Genome 430 2.0 Array 1429452_x_at - -
          Mouse Genome 430 2.0 Array 1453144_at - -
          Mouse Genome 430 2.0 Array 1453144_PM_at - -
          Mouse Genome 430 2.0 Array 1429452_PM_x_at - -
          Mouse Genome 430 2.0 Array 1453145_PM_at - -
          Mouse Genome 430 2.0 Array 1453145_at - -
          Mouse Genome 430 2.0 Array 1434975_x_at - -
          Mouse Genome 430 2.0 Array 1434975_PM_x_at - -
          Mouse Genome 430 2.0 Array 1439069_a_at - -
          Mouse Expression Set 430 1426387_x_at - -
          Mouse Expression Set 430 1436944_PM_x_at - -
          Mouse Expression Set 430 1426387_PM_x_at - -
          Mouse Expression Set 430 1435353_PM_a_at - -
          Mouse Expression Set 430 1439069_a_at - -
          Mouse Expression Set 430 1436944_x_at - -
          Mouse Expression Set 430 1439070_x_at - -
          Mouse Expression Set 430 1434975_PM_x_at - -
          Mouse Expression Set 430 1435353_a_at - -
          Mouse Expression Set 430 1439070_PM_x_at - -
          Mouse Expression Set 430 1439069_PM_a_at - -
          Mouse Expression Set 430 1434975_x_at - -
          Mouse Expression Set 430 1453145_at - -
          Mouse Expression Set 430 1429452_PM_x_at - -
          Mouse Expression Set 430 1454566_at - -
          Mouse Expression Set 430 1435426_s_at - -
          Mouse Expression Set 430 1435426_PM_s_at - -
          Mouse Expression Set 430 1435425_PM_at - -
          Mouse Expression Set 430 1453145_PM_at - -
          Mouse Expression Set 430 1435425_at - -
          Mouse Expression Set 430 1454566_PM_at - -
          Mouse Expression Set 430 1453144_at - -
          Mouse Expression Set 430 1453144_PM_at - -
          Mouse Expression Set 430 1429452_x_at - -
          Murine Genome U74v2 Set 97448_PM_at - -
          Murine Genome U74v2 Set 97448_at - -
          Murine Genome U74v2 Set 165802_r_at - -
          Murine Genome U74v2 Set 165802_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1439070_x_at - -
          Mouse Genome 430A 2.0 Array 1439069_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1434975_x_at - -
          Mouse Genome 430A 2.0 Array 1434975_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1435353_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1439069_a_at - -
          Mouse Genome 430A 2.0 Array 1436944_x_at - -
          Mouse Genome 430A 2.0 Array 1426387_x_at - -
          Mouse Genome 430A 2.0 Array 1439070_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1435353_a_at - -
          Mouse Genome 430A 2.0 Array 1426387_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1436944_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1434975_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1439069_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1434975_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1439069_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1453144_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453145_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453145_at - -
          Affymetrix HT Mouse Genome 430B Array 1453144_at - -
          Affymetrix HT Mouse Genome 430B Array 1429452_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1429452_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 106980129 - -
          Illumina mouse-6 v1.0 expression beadchip 4780010 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004780010 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2477946 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2477946 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252359 - -
          mouse-6 v1.1 (Illumina) scl0001464.1_61-S - -
          Illumina Sentrix mouse-6.v1 bead array 4780010 - -
          Illumina Sentrix mouse-6.v1 bead array 106980129 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001464.1_61-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4780010 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106980129 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2543630 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2477946 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001464.1_61-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0