Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600009284.mm.1 |
TC0600001203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009284.mm.1 |
NM_011498.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009284.mm.1 |
NM_011498.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009283.mm.1 |
TC0600001203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009283.mm.1 |
NM_011498.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009283.mm.1 |
NM_011498.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009280.mm.1 |
TC0600001203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009280.mm.1 |
NM_011498.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009280.mm.1 |
NM_011498.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009281.mm.1 |
TC0600001203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009281.mm.1 |
NM_011498.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009281.mm.1 |
NM_011498.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009282.mm.1 |
TC0600001203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009282.mm.1 |
NM_011498.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009282.mm.1 |
NM_011498.4 |
Refseq |