Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01183111_m1
          See other FGF6 GE Assays ›
          Gene Symbol:
          Fgf6
          Gene Name:
          fibroblast growth factor 6
          Gene Aliases:
          Fgf-6, HSTF-2
          Chromosome Location:
          Chr.6: 127015542 - 127028182 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01183111_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgf6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010204.1 NP_034334.1 2 - 3 500 111
          GenBank mRNA AK086530.1 - 2 - 3 418 111
          M92416.1 - 2 - 3 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001475.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          angiogenesis
          multicellular organism development
          positive regulation of cell population proliferation
          fibroblast growth factor receptor signaling pathway
          cell differentiation
          myoblast differentiation
          positive regulation of cell division
          signaling receptor binding
          fibroblast growth factor receptor binding
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073612 NM_010204.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073612 AK086530.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073612 M92416.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 846808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906832 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137931 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133188 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145057 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073612 NM_010204.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073612 AK086530.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073612 M92416.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P155536 NM_010204.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P155536 AK086530.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P155536 M92416.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P351039 M92416.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133188 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145057 - -
          Whole Mouse Genome G4122A A_51_P351039 M92416.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P155536 NM_010204.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P155536 AK086530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P155536 M92416.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145057 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05231917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05424161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05872591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05818172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05295393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05463802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05463811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05333482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05379308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05295394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05818177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05695334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05231922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05872596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05445466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05231916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05055009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05695332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05463821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05872594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24830198 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542026 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542025 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542024 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542027 - -
          Affymetrix HT MG-430 PM Array Plate 1427582_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10542023 - -
          Mouse Genome 430 2.0 Array 1427582_PM_at - -
          Mouse Genome 430 2.0 Array 1427582_at - -
          Affymetrix Murine Genome 6500A Array Msa14200_at - -
          Affymetrix Mouse Clariom S Array PSR0600012171.mm.1 TC0600001475.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012171.mm.1 NM_010204.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012171.mm.1 NM_010204.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012170.mm.1 TC0600001475.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012170.mm.1 NM_010204.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012170.mm.1 NM_010204.1 Refseq
          Mouse Expression Set 430 1427582_at - -
          Mouse Expression Set 430 1427582_PM_at - -
          Murine Genome U74v2 Set 97121_PM_at - -
          Murine Genome U74v2 Set 97121_at - -
          Mouse Genome 430A 2.0 Array 1427582_PM_at - -
          Mouse Genome 430A 2.0 Array 1427582_at - -
          Affymetrix HT Mouse Genome 430A Array 1427582_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427582_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 616261 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218948 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2613997 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2613997 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2958386 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2958386 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0