Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01184322_m1
          See other PPARG GE Assays ›
          Gene Symbol:
          Pparg
          Gene Name:
          peroxisome proliferator activated receptor gamma
          Gene Aliases:
          Nr1c3, PPAR-gamma, PPAR-gamma2, PPARgamma, PPARgamma2
          Chromosome Location:
          Chr.6: 115360879 - 115490404 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01184322_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pparg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011146.3 NP_035276.2 4 - 5 669 101
          GenBank mRNA AB644275.1 - 5 - 6 675 101
          AY208183.1 - 4 - 5 669 101
          AY208184.1 - 4 - 5 669 101
          AY243585.1 - 4 - 5 669 101
          BC021798.1 - 4 - 5 669 4191539 101
          CT010340.1 - 4 - 5 624 101
          EF062476.1 - 4 - 5 660 101
          GQ868647.1 - 3 - 4 427 101
          GQ868648.1 - 3 - 4 407 101
          GQ868649.1 - 3 - 4 427 101
          GQ868650.1 - 3 - 4 407 101
          U01664.1 - 5 - 6 701 101
          U01841.1 - 5 - 6 1024 101
          U09138.1 - 4 - 5 663 101
          U10374.1 - 3 - 4 534 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001279.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Mammalian Gene Collection Mus musculus Mouse lipid regulated genes Inventoried Mouse_lipid_regulated_genes Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pparg

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          negative regulation of cytokine production
          placenta development
          response to dietary excess
          diet induced thermogenesis
          negative regulation of acute inflammatory response
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory response
          signal transduction
          regulation of blood pressure
          negative regulation of cell proliferation
          response to light stimulus
          regulation of gene expression
          negative regulation of macrophage derived foam cell differentiation
          negative regulation of receptor biosynthetic process
          negative regulation of cholesterol storage
          negative regulation of sequestering of triglyceride
          long-chain fatty acid transport
          regulation of lipid metabolic process
          fatty acid oxidation
          monocyte differentiation
          negative regulation of cell growth
          intracellular receptor signaling pathway
          epithelial cell differentiation
          response to food
          response to retinoic acid
          cellular response to insulin stimulus
          negative regulation of collagen biosynthetic process
          response to lipid
          peroxisome proliferator activated receptor signaling pathway
          glucose homeostasis
          regulation of circadian rhythm
          lipoprotein transport
          positive regulation of apoptotic process
          steroid hormone mediated signaling pathway
          cell fate commitment
          fat cell differentiation
          regulation of fat cell differentiation
          positive regulation of fat cell differentiation
          low-density lipoprotein particle receptor biosynthetic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fatty acid oxidation
          cell maturation
          rhythmic process
          negative regulation of smooth muscle cell proliferation
          positive regulation of oligodendrocyte differentiation
          white fat cell differentiation
          brown fat cell differentiation
          positive regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of telomerase activity
          response to low-density lipoprotein particle
          positive regulation of phagocytosis, engulfment
          negative regulation of interferon-gamma-mediated signaling pathway
          regulation of transcription involved in cell fate commitment
          cellular response to lithium ion
          cellular response to organic cyclic compound
          negative regulation of peptide hormone secretion
          negative regulation of cellular response to insulin stimulus
          negative regulation of pancreatic stellate cell proliferation
          RNA polymerase II regulatory region DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          transcription factor binding
          drug binding
          zinc ion binding
          enzyme binding
          protein phosphatase binding
          estrogen receptor binding
          ligand-dependent nuclear receptor transcription coactivator activity
          activating transcription factor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          metal ion binding
          retinoid X receptor binding
          arachidonic acid binding
          WW domain binding
          alpha-actinin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600010097.mm.1 TC0600001279.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600010097.mm.1 NM_001127330.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010097.mm.1 NM_011146.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010097.mm.1 NM_011146.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0