Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083922.1 | NP_001077391.1 | 2 - 3 | 364 | 72 | |
NM_016757.2 | NP_058037.2 | 2 - 3 | 367 | 72 | ||
GenBank mRNA | AK076948.1 | - | 2 - 3 | 299 | 72 | |
AK152822.1 | - | 2 - 3 | 369 | 72 | ||
AK178563.1 | - | 1 - 2 | 101 | 72 | ||
AK179139.1 | - | 2 - 3 | 220 | 72 | ||
BC005666.1 | - | 2 - 3 | 483 | 3152660 | 72 | |
CX205200.1 | - | 2 - 3 | 338 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |