Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01184921_g1
          See other WBP1 GE Assays ›
          Gene Symbol:
          Wbp1
          Gene Name:
          WW domain binding protein 1
          Gene Aliases:
          AI255152
          Chromosome Location:
          Chr.6: 83119044 - 83121663 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01184921_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001083922.1 NP_001077391.1 4 - 5 643 81
          NM_001083923.1 NP_001077392.1 3 - 4 541 81
          NM_016757.2 NP_058037.2 4 - 5 646 81
          GenBank mRNA AK076948.1 - 4 - 5 578 81
          BC005666.1 - 4 - 5 762 3152660 81
          BC019889.1 - 3 - 4 465 5123630 81
          BE860581.1 - 3 - 4 375 81
          U40825.1 - 4 - 5 608 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002560.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wbp1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein binding
          WW domain binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160372 BC019889.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160372 AK190643.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160372 BC005666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160372 U40825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160372 AK170907.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160372 BC019889.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160372 AK190643.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160372 BC005666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160372 U40825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160372 AK170907.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11541 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117956 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160372 BC019889.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160372 AK190643.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160372 BC005666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160372 U40825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160372 AK170907.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160372 BC019889.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160372 AK190643.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160372 BC005666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160372 U40825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160372 AK170907.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160372 BC019889.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160372 AK190643.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160372 BC005666.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160372 U40825.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160372 AK170907.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160372 BC019889.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160372 AK190643.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160372 BC005666.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160372 U40825.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160372 AK170907.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591549 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117956 - -
          Whole Mouse Genome G4122A A_51_P160372 BC019889.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160372 AK190643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160372 BC005666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160372 U40825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160372 AK170907.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160372 BC019889.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160372 AK190643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160372 BC005666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160372 U40825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160372 AK170907.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05303346 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05276721 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05276723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05303292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05303331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05303335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05276723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05303320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591554 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545649 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545648 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545650 - -
          Affymetrix HT MG-430 PM Array Plate 1449441_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545644 - -
          Mouse Genome 430 2.0 Array 1449441_PM_a_at - -
          Mouse Genome 430 2.0 Array 1449441_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22377_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22377_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600019832.mm.1 TC0600002560.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019832.mm.1 NM_001083922.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019832.mm.1 NM_001083922.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019832.mm.1 NM_001083923.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019832.mm.1 NM_001083923.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019832.mm.1 NM_016757.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019832.mm.1 NM_016757.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019835.mm.1 TC0600002560.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019835.mm.1 NM_001083922.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019835.mm.1 NM_001083922.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019835.mm.1 NM_001083923.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019835.mm.1 NM_001083923.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019835.mm.1 NM_016757.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019835.mm.1 NM_016757.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019837.mm.1 TC0600002560.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019837.mm.1 NM_001083922.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019837.mm.1 NM_001083922.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019837.mm.1 NM_001083923.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019837.mm.1 NM_001083923.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019837.mm.1 NM_016757.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019837.mm.1 NM_016757.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019831.mm.1 TC0600002560.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019831.mm.1 NM_001083922.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019831.mm.1 NM_001083922.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019831.mm.1 NM_001083923.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019831.mm.1 NM_001083923.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019831.mm.1 NM_016757.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019831.mm.1 NM_016757.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019830.mm.1 TC0600002560.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019830.mm.1 NM_001083922.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019830.mm.1 NM_001083922.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019830.mm.1 NM_001083923.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019830.mm.1 NM_001083923.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019830.mm.1 NM_016757.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019830.mm.1 NM_016757.2 Refseq
          Mouse Expression Set 430 1449441_PM_a_at - -
          Mouse Expression Set 430 1449441_a_at - -
          Murine Genome U74v2 Set 92619_PM_at - -
          Murine Genome U74v2 Set 92619_at - -
          Mouse Genome 430A 2.0 Array 1449441_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1449441_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449441_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449441_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6100066 - -
          Illumina mouse-6 v1.0 expression beadchip 6100066 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006100066 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2454788 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2454788 - -
          mouse-6 v1.1 (Illumina) scl28804.3_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 6100066 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28804.3_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100066 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2454788 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2951120 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2951120 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2454788 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix