Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01185044_g1
          See other EMG1 GE Assays ›
          Gene Symbol:
          Emg1
          Gene Name:
          EMG1 nucleolar protein homolog (S. cerevisiae)
          Gene Aliases:
          C2f, Grcc2f
          Chromosome Location:
          Chr.6: 124704370 - 124712178 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01185044_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Emg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013536.2 NP_038564.1 5 - 6 757 102
          GenBank mRNA AK081757.1 - 5 - 6 735 102
          AK218607.1 - 3 - 4 278 102
          BC002004.1 - 5 - 6 677 3487480 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003110.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Emg1

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst development
          rRNA processing
          nucleologenesis
          rRNA methylation
          methylation
          ribosome biogenesis
          rRNA base methylation
          rRNA (adenine-N6,N6-)-dimethyltransferase activity
          RNA binding
          protein C-terminal carboxyl O-methyltransferase activity
          tRNA (guanine-N2-)-methyltransferase activity
          methyltransferase activity
          C-methyltransferase activity
          N-methyltransferase activity
          O-methyltransferase activity
          S-methyltransferase activity
          RNA methyltransferase activity
          mRNA methyltransferase activity
          tRNA methyltransferase activity
          protein methyltransferase activity
          2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
          rRNA methyltransferase activity
          rRNA (uridine-2'-O-)-methyltransferase activity
          rRNA (adenine-N6-)-methyltransferase activity
          DNA-methyltransferase activity
          tRNA (guanine-N1-)-methyltransferase activity
          rRNA (cytosine-C5-)-methyltransferase activity
          selenocysteine methyltransferase activity
          arginine N-methyltransferase activity
          protein-arginine N-methyltransferase activity
          lysine N-methyltransferase activity
          protein-lysine N-methyltransferase activity
          tRNA (uracil) methyltransferase activity
          tRNA (guanine) methyltransferase activity
          tRNA (adenine) methyltransferase activity
          tRNA (cytosine) methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity
          tRNA (adenine-N1-)-methyltransferase activity
          rRNA (adenine) methyltransferase activity
          rRNA (cytosine) methyltransferase activity
          rRNA (guanine) methyltransferase activity
          rRNA (uridine) methyltransferase activity
          transferase activity
          protein C-terminal leucine carboxyl O-methyltransferase activity
          1-phenanthrol methyltransferase activity
          protein-arginine N5-methyltransferase activity
          rRNA binding
          methylarsonite methyltransferase activity
          dimethylarsinite methyltransferase activity
          4,5-dihydroxybenzo(a)pyrene methyltransferase activity
          1-hydroxypyrene methyltransferase activity
          1-hydroxy-6-methoxypyrene methyltransferase activity
          demethylmenaquinone methyltransferase activity
          cobalt-precorrin-6B C5-methyltransferase activity
          cobalt-precorrin-7 C15-methyltransferase activity
          cobalt-precorrin-5B C1-methyltransferase activity
          cobalt-precorrin-3 C17-methyltransferase activity
          dimethylamine methyltransferase activity
          hydroxyneurosporene-O-methyltransferase activity
          tRNA (adenine-57, 58-N(1)-) methyltransferase activity
          methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
          trimethylamine methyltransferase activity
          methanol-specific methylcobalamin:coenzyme M methyltransferase activity
          monomethylamine methyltransferase activity
          P-methyltransferase activity
          Se-methyltransferase activity
          2-phytyl-1,4-naphthoquinone methyltransferase activity
          tRNA (uracil-2'-O-)-methyltransferase activity
          tRNA (cytosine-2'-O-)-methyltransferase activity
          phosphomethylethanolamine N-methyltransferase activity
          tRNA (cytosine-3-)-methyltransferase activity
          rRNA (pseudouridine) methyltransferase activity
          rRNA (cytosine-2'-O-)-methyltransferase activity
          rRNA (cytosine-N4-)-methyltransferase activity
          trihydroxyferuloyl spermidine O-methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816528 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816526 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518586 BC002004.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518586 AK218607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518586 NM_013536.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518586 AK081757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518586 BC002004.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518586 AK218607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518586 NM_013536.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518586 AK081757.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300195 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 113231 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127052 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518586 BC002004.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518586 AK218607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518586 NM_013536.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518586 AK081757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518586 BC002004.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518586 AK218607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518586 NM_013536.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518586 AK081757.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518586 BC002004.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518586 AK218607.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518586 NM_013536.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518586 AK081757.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518586 BC002004.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518586 AK218607.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518586 NM_013536.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518586 AK081757.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816524 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127052 - -
          Whole Mouse Genome G4122A A_51_P518586 BC002004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518586 AK218607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518586 NM_013536.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518586 AK081757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518586 BC002004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518586 AK218607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518586 NM_013536.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518586 AK081757.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112083 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127052 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006639 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006653 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006636 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816528 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816526 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05006636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05659509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05006653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05443279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05659480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816504 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10547762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547764 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547760 - -
          Affymetrix HT MG-430 PM Array Plate 1416393_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10547758 - -
          Mouse Genome 430 2.0 Array 1416393_at - -
          Mouse Genome 430 2.0 Array 1416393_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14791_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14791_at - -
          Affymetrix Mouse Clariom S Array PSR0600024048.mm.1 TC0600003110.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024048.mm.1 NM_013536.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024048.mm.1 NM_013536.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024047.mm.1 TC0600003110.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024047.mm.1 NM_013536.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024047.mm.1 NM_013536.2 Refseq
          Mouse Expression Set 430 1416393_at - -
          Mouse Expression Set 430 1416393_PM_at - -
          Murine Genome U74v2 Set 160531_PM_at - -
          Murine Genome U74v2 Set 160531_at - -
          Mouse Genome 430A 2.0 Array 1416393_PM_at - -
          Mouse Genome 430A 2.0 Array 1416393_at - -
          Affymetrix HT Mouse Genome 430A Array 1416393_at - -
          Affymetrix HT Mouse Genome 430A Array 1416393_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 380242 - -
          Illumina mouse-6 v1.0 expression beadchip 50286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000380242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000050286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2744237 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2735981 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2744237 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2735981 - -
          mouse-6 v1.1 (Illumina) scl0001050.1_29-S - -
          mouse-6 v1.1 (Illumina) scl28426.4.1_40-S - -
          Illumina Sentrix mouse-6.v1 bead array 380242 - -
          Illumina Sentrix mouse-6.v1 bead array 50286 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28426.4.1_40-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001050.1_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 50286 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 380242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2735981 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2744237 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2744237 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2735981 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001050.1_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl28426.4.1_40-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0