Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01185105_g1
          See other PHB2 GE Assays ›
          Gene Symbol:
          Phb2
          Gene Name:
          prohibitin 2
          Gene Aliases:
          AU044498, BAP, Bap37, Bcap37, REA
          Chromosome Location:
          Chr.6: 124712289 - 124716945 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01185105_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007531.2 NP_031557.2 7 - 8 940 60
          GenBank mRNA AK002334.1 - 7 - 8 906 60
          AK011107.1 - 7 - 8 907 60
          AK090088.1 - 7 - 8 897 60
          AK132052.1 - 7 - 8 902 60
          AK132058.1 - 7 - 8 929 60
          AK135729.1 - 7 - 8 857 60
          AK151952.1 - 7 - 8 934 60
          AK152375.1 - 6 - 7 1589 60
          AK153368.1 - 6 - 7 1589 60
          AY211613.1 - 7 - 8 902 60
          AY319418.1 - 7 - 8 792 60
          BC145875.1 - 7 - 8 892 40131178 60
          X78683.1 - 7 - 8 896 60

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Phb2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mitochondrion organization
          sister chromatid cohesion
          positive regulation of exit from mitosis
          negative regulation of intracellular estrogen receptor signaling pathway
          negative regulation of mammary gland epithelial cell proliferation
          negative regulation of apoptotic process
          negative regulation of DNA-binding transcription factor activity
          negative regulation of transcription, DNA-templated
          protein stabilization
          positive regulation of DNA-binding transcription factor activity
          mammary gland branching involved in thelarche
          mammary gland alveolus development
          regulation of branching involved in mammary gland duct morphogenesis
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of cell cycle G1/S phase transition
          chromatin binding
          protein binding
          protein C-terminus binding
          amide binding
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816534 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816535 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816532 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816531 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816533 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126523 X78683.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126523 BC145875.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126523 AK153368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126523 AY211613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126523 AY319418.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157019 X78683.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157019 BC145875.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157019 AK153368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157019 AY211613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157019 AY319418.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908330 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126523 X78683.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126523 BC145875.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126523 AK153368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126523 AY211613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126523 AY319418.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157019 X78683.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157019 BC145875.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157019 AK153368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157019 AY211613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157019 AY319418.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P205833 X78683.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P205833 BC145875.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P205833 AY211613.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P205833 AK151952.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P205833 AK132058.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816568 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06056139 - -
          Whole Mouse Genome G4122A A_51_P205833 X78683.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205833 BC145875.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205833 AY211613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205833 AK151952.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205833 AK132058.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101672 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126523 X78683.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126523 BC145875.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126523 AK153368.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126523 AY211613.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126523 AY319418.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05492562 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05492541 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006657 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05492543 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006655 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05492527 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05492507 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05492536 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816531 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816534 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816532 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816533 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05492529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05006654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05006647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05492547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05009578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816556 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10541720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541715 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541718 - -
          Affymetrix HT MG-430 PM Array Plate 1460035_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455713_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1416202_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438938_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10541711 - -
          Mouse Genome 430 2.0 Array 1416202_PM_at - -
          Mouse Genome 430 2.0 Array 1460035_at - -
          Mouse Genome 430 2.0 Array 1438938_x_at - -
          Mouse Genome 430 2.0 Array 1455713_PM_x_at - -
          Mouse Genome 430 2.0 Array 1455713_x_at - -
          Mouse Genome 430 2.0 Array 1438938_PM_x_at - -
          Mouse Genome 430 2.0 Array 1460035_PM_at - -
          Mouse Genome 430 2.0 Array 1416202_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12034_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12034_at - -
          Mouse Expression Set 430 1455713_x_at - -
          Mouse Expression Set 430 1438938_PM_x_at - -
          Mouse Expression Set 430 1416202_PM_at - -
          Mouse Expression Set 430 1455713_PM_x_at - -
          Mouse Expression Set 430 1416202_at - -
          Mouse Expression Set 430 1438938_x_at - -
          Mouse Expression Set 430 1460035_PM_at - -
          Mouse Expression Set 430 1460035_at - -
          Murine Genome U74v2 Set 98545_PM_at - -
          Murine Genome U74v2 Set 98545_at - -
          Mouse Genome 430A 2.0 Array 1416202_PM_at - -
          Mouse Genome 430A 2.0 Array 1438938_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1416202_at - -
          Mouse Genome 430A 2.0 Array 1438938_x_at - -
          Mouse Genome 430A 2.0 Array 1455713_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1455713_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438938_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1416202_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455713_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438938_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1455713_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1416202_at - -
          Affymetrix HT Mouse Genome 430B Array 1460035_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460035_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1230050 - -
          Illumina mouse-6 v1.0 expression beadchip 1230050 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001230050 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224868 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224868 - -
          mouse-6 v1.1 (Illumina) scl29519.10.9_31-S - -
          Illumina Sentrix mouse-6.v1 bead array 1230050 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29519.10.9_31-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1230050 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1224868 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224868 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl29519.10.9_31-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline