Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01185617_g1
          See other 4930591E09RIK GE Assays ›
          Gene Symbol:
          4930591E09Rik
          Gene Name:
          RIKEN cDNA 4930591E09 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 38175774 - 38181903 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01185617_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930591E09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016383.1 - 2 - 3 327 65
          BY716035.1 - 2 - 3 328 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          26535
          Gm26535 -801.7kb Sense Chr. 10 - 37319505 - 37379652 on NCBI Build GRCm38
          Downstream ret finger protein-like 4B Rfpl4b 100040915,
          Gm3037,
          Gm62
          634.8kb Anti-sense Chr. 10 - 38816682 - 38822938 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139232 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08766321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27073130 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 662690 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0