Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01185788_g1
          See other NKAIN1 GE Assays ›
          Gene Symbol:
          Nkain1
          Gene Name:
          Na+/K+ transporting ATPase interacting 1
          Gene Aliases:
          2610200G18Rik, 2810426C15Rik
          Chromosome Location:
          Chr.4: 130530131 - 130574036 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01185788_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nkain1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025998.3 NP_080274.2 6 - 7 772 87
          GenBank mRNA AK011164.1 - 6 - 7 905 87
          AK011850.1 - 6 - 7 703 87
          EF058048.2 - 6 - 7 825 87

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nkain1

          Gene Ontology Categories:

          Process(es)

          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P670978 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025368 CV563214.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025368 AK160181.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025368 NM_025998.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025368 AK009389.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 314986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509056 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 AF175771.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 AF291425.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 AJ279791.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 AJ279792.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 AJ279793.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 AJ279794.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 AJ279795.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 AK013248.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 AK154694.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 AK171199.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 AY039000.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 BC051204.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 BC055858.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599856 NM_020286.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707681 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 46938 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719477 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 479011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434943 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632161 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290243 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 857994 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135599 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025368 CV563214.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025368 AK160181.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025368 NM_025998.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025368 AK009389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P670978 - -
          Whole Mouse Genome G4122A A_51_P244092 AK218646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P244092 AK218997.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P244092 AK217347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P244092 AK218211.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P244092 BC116391.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P670978 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116239 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136206 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608709 - -
          Affymetrix HT MG-430 PM Array Plate 1449553_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433274_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419652_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1419651_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433273_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608709 - -
          Mouse Genome 430 2.0 Array 1419652_s_at - -
          Mouse Genome 430 2.0 Array 1449553_PM_at - -
          Mouse Genome 430 2.0 Array 1433273_PM_at - -
          Mouse Genome 430 2.0 Array 1419652_PM_s_at - -
          Mouse Genome 430 2.0 Array 1419651_at - -
          Mouse Genome 430 2.0 Array 1433273_at - -
          Mouse Genome 430 2.0 Array 1433274_PM_at - -
          Mouse Genome 430 2.0 Array 1433274_at - -
          Mouse Genome 430 2.0 Array 1419651_PM_at - -
          Mouse Genome 430 2.0 Array 1449553_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67149_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67149_at - -
          Mouse Expression Set 430 1449553_PM_at - -
          Mouse Expression Set 430 1419652_PM_s_at - -
          Mouse Expression Set 430 1419651_at - -
          Mouse Expression Set 430 1419651_PM_at - -
          Mouse Expression Set 430 1419652_s_at - -
          Mouse Expression Set 430 1449553_at - -
          Murine Genome U74v2 Set 162249_PM_f_at - -
          Murine Genome U74v2 Set 104229_PM_at - -
          Murine Genome U74v2 Set 104229_at - -
          Murine Genome U74v2 Set 162249_f_at - -
          Mouse Genome 430A 2.0 Array 1419651_PM_at - -
          Mouse Genome 430A 2.0 Array 1449553_at - -
          Mouse Genome 430A 2.0 Array 1419652_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1419652_s_at - -
          Mouse Genome 430A 2.0 Array 1449553_PM_at - -
          Mouse Genome 430A 2.0 Array 1419651_at - -
          Affymetrix HT Mouse Genome 430A Array 1419651_at - -
          Affymetrix HT Mouse Genome 430A Array 1449553_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419651_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449553_at - -
          Affymetrix HT Mouse Genome 430A Array 1419652_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419652_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1433273_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433273_at - -
          Affymetrix HT Mouse Genome 430B Array 1433274_at - -
          Affymetrix HT Mouse Genome 430B Array 1433274_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6510692 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006510692 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605041 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605041 - -
          mouse-6 v1.1 (Illumina) scl24894.7.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 6510692 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24894.7.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6510692 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605041 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605041 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24894.7.1_6-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0