Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01187143_m1
          See other 4933407E24RIK GE Assays ›
          Gene Symbol:
          4933407E24Rik
          Gene Name:
          RIKEN cDNA 4933407E24 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 124569179 - 124575538 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01187143_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933407E24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045819.1 2 - 3 402 76
          GenBank mRNA AK160260.1 - 2 - 3 391 76
          BB616969.1 - 2 - 3 391 76
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB615438.1 - 2 - 3 387 Chr. 4 - 124569195 - 124574414 [+] on Build GRCm38 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 2164 Gm2164 Anti-sense Chr. 4 - 124522925 - 124657504 on NCBI Build GRCm38
          Downstream POU domain,
          class 3,
          transcription factor 1
          Pou3f1 Oct-6,
          Oct6,
          Otf-6,
          Otf6,
          Scip,
          Test1,
          Tst-1,
          Tst1
          83.1kb Sense Chr. 4 - 124657646 - 124660655 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001416.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK160260.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126104 AK160260.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125917 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126104 AK160260.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125917 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03375885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03584495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03593764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03992703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03375892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03699892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03644634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03992710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03992707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03850294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03992709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03883802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03644644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03482955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03542674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03992687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03542676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03483811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03992685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04013062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03992696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03644632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03644636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03243853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03353727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03992694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03883799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03883798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03375888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03811922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03811924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03644625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03644645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03937337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03567737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03992715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03310793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23268931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269035 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400010648.mm.1 TC0400001416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400010648.mm.1 NR_045819.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010648.mm.1 NR_045819.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010646.mm.1 TC0400001416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400010646.mm.1 NR_045819.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010646.mm.1 NR_045819.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010647.mm.1 TC0400001416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400010647.mm.1 NR_045819.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010647.mm.1 NR_045819.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010649.mm.1 TC0400001416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400010649.mm.1 NR_045819.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010649.mm.1 NR_045819.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 829464 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0