Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01188456_g1
          See other IQCC GE Assays ›
          Gene Symbol:
          Iqcc
          Gene Name:
          IQ motif containing C
          Gene Aliases:
          BC026978
          Chromosome Location:
          Chr.4: 129615617 - 129619213 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01188456_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Iqcc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198026.2 NP_932143.2 4 - 5 575 82
          GenBank mRNA BC026978.1 - 2 - 3 350 82
          BC031493.1 - 552 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Iqcc

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23297599 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23297598 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23297601 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23297597 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23297600 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P371063 BY123003.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P371063 AK083217.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P371063 NM_198026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P371063 AK045767.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P371063 BY123003.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P371063 AK083217.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P371063 NM_198026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P371063 AK045767.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158458 NM_198026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158458 BC026978.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909794 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170956 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158458 NM_198026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158458 BC026978.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966714 BC031493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P371063 BY123003.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P371063 AK083217.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P371063 NM_198026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P371063 AK045767.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P371063 BY123003.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P371063 AK083217.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P371063 NM_198026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P371063 AK045767.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23297594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23297589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23297584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23297591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23297598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23297585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23297592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23297588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23297586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23297583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23297596 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170956 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05400095 - -
          Whole Mouse Genome G4122A A_52_P364279 BC031493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P364279 NM_198026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P364279 BC026978.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P371063 BY123003.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P371063 AK083217.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P371063 NM_198026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P371063 AK045767.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P371063 BY123003.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P371063 AK083217.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P371063 NM_198026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P371063 AK045767.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118983 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03981846 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03981836 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03981837 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23297599 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23297601 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23297597 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23297598 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23297600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23297528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23297559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03874332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03981838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03981846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04010025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23297577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03724666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23297592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23297564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03874329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03981842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03811895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23297569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03874325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23297561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23297591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03811899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03874328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23297598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03874326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23297576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23297589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03874331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04077427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03874322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23297575 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10516657 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516654 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516656 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516653 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516655 - -
          Affymetrix HT MG-430 PM Array Plate 1452356_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10516652 - -
          Mouse Genome 430 2.0 Array 1452356_at - -
          Mouse Genome 430 2.0 Array 1452356_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230767_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230767_at - -
          Affymetrix Mouse Clariom S Array PSR0400029430.mm.1 TC0400003606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029430.mm.1 NM_198026.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029430.mm.1 NM_198026.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029432.mm.1 TC0400003606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029432.mm.1 NM_198026.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029432.mm.1 NM_198026.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029429.mm.1 TC0400003606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029429.mm.1 NM_198026.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029429.mm.1 NM_198026.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029431.mm.1 TC0400003606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029431.mm.1 NM_198026.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029431.mm.1 NM_198026.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029433.mm.1 TC0400003606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029433.mm.1 NM_198026.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029433.mm.1 NM_198026.2 Refseq
          Mouse Expression Set 430 1452356_at - -
          Mouse Expression Set 430 1452356_PM_at - -
          Murine Genome U74v2 Set 139246_at - -
          Murine Genome U74v2 Set 139246_PM_at - -
          Mouse Genome 430A 2.0 Array 1452356_PM_at - -
          Mouse Genome 430A 2.0 Array 1452356_at - -
          Affymetrix HT Mouse Genome 430A Array 1452356_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452356_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4010484 - -
          Illumina mouse-6 v1.0 expression beadchip 4010484 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004010484 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769172 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769172 - -
          mouse-6 v1.1 (Illumina) scl23762.4_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 4010484 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23762.4_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4010484 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2769171 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2769171 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl23762.4_16-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0