Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000002385.mm.1 |
TC1000000358.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002385.mm.1 |
NM_031877.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002385.mm.1 |
NM_031877.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002387.mm.1 |
TC1000000358.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002387.mm.1 |
NM_031877.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002387.mm.1 |
NM_031877.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002389.mm.1 |
TC1000000358.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002389.mm.1 |
NM_031877.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002389.mm.1 |
NM_031877.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002390.mm.1 |
TC1000000358.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002390.mm.1 |
NM_031877.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002390.mm.1 |
NM_031877.3 |
Refseq |