Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01190364_m1
          See other DUSP11 GE Assays ›
          Gene Symbol:
          Dusp11
          Gene Name:
          dual specificity phosphatase 11 (RNA/RNP complex 1-interacting)
          Gene Aliases:
          2010300F21Rik, AI481497
          Chromosome Location:
          Chr.6: 85942268 - 85962045 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01190364_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dusp11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028099.4 NP_082375.4 1 - 2 235 65
          GenBank mRNA AK040232.1 - 1 - 2 213 65
          AK081629.1 - 1 - 2 212 65
          AK088941.1 - 1 - 2 232 65
          BB613140.1 - 1 - 2 236 65
          BC067028.1 - 1 - 2 189 30364073 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002629.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dusp11

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          RNA metabolic process
          dephosphorylation
          polynucleotide 5' dephosphorylation
          RNA binding
          polynucleotide 5'-phosphatase activity
          protein tyrosine phosphatase activity
          protein tyrosine/serine/threonine phosphatase activity
          protein tyrosine/threonine phosphatase activity
          JUN kinase phosphatase activity
          inositol bisphosphate phosphatase activity
          hydrolase activity
          phosphatase activity
          MAP kinase tyrosine/serine/threonine phosphatase activity
          inositol-1,3,4-trisphosphate 4-phosphatase activity
          NADP phosphatase activity
          transmembrane receptor protein phosphatase activity
          inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
          inositol-1,4,5,6-tetrakisphosphate 6-phosphatase activity
          inositol-4,5-bisphosphate 5-phosphatase activity
          5-amino-6-(5-phosphoribitylamino)uracil phosphatase activity
          phosphatidylinositol-4-phosphate phosphatase activity
          phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
          phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
          inositol phosphate phosphatase activity
          inositol-1,3,4,5,6-pentakisphosphate 1-phosphatase activity
          inositol-3,4-bisphosphate 4-phosphatase activity
          inositol-1,3,4-trisphosphate 1-phosphatase activity
          inositol-1,3,4,6-tetrakisphosphate 6-phosphatase activity
          inositol-1,3,4,6-tetrakisphosphate 1-phosphatase activity
          phosphatidylinositol-1,4,5-trisphosphate 5-phosphatase activity
          nucleotide phosphatase activity, acting on free nucleotides
          protein histidine phosphatase activity
          inosine-diphosphatase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600020425.mm.1 TC0600002629.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020425.mm.1 NM_028099.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020425.mm.1 NM_028099.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020428.mm.1 TC0600002629.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020428.mm.1 NM_028099.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020428.mm.1 NM_028099.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020423.mm.1 TC0600002629.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020423.mm.1 NM_028099.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020423.mm.1 NM_028099.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6bbccd986b-mx52t:80/100.66.130.62:80.
          git-commit: 717389b07c2b9f75290ff02e811d41bb86d4254a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0