Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01190872_g1
          See other 1700069P05RIK GE Assays ›
          Gene Symbol:
          1700069P05Rik
          Gene Name:
          RIKEN cDNA 1700069P05 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 118246695 - 118248454 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01190872_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700069P05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040527.1 2 - 3 660 54
          GenBank mRNA AK006926.1 - 2 - 3 664 54

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ret proto-oncogene Ret PTC,
          RET51,
          RET9,
          c-Ret
          -49.0kb Sense Chr. 6 - 118151748 - 118197744 on NCBI Build GRCm38
          Downstream predicted gene,
          30498
          Gm30498 52.8kb Sense Chr. 6 - 118300167 - 118308719 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002997.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1147358 AK006926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1147358 AK006926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1147358 AK006926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1147358 AK006926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1147358 AK006926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1147358 AK006926.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24785750 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600023039.mm.1 TC0600002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600023039.mm.1 NR_040527.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600023039.mm.1 NR_040527.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600023038.mm.1 TC0600002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600023038.mm.1 NR_040527.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600023038.mm.1 NR_040527.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600023037.mm.1 TC0600002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600023037.mm.1 NR_040527.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600023037.mm.1 NR_040527.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 618670 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline