Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01190908_m1
          See other 4930444F02RIK GE Assays ›
          Gene Symbol:
          4930444F02Rik
          Gene Name:
          RIKEN cDNA 4930444F02 gene
          Gene Aliases:
          1700010L24Rik, mpiR
          Chromosome Location:
          Chr.10: 18797299 - 18831930 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01190908_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930444F02Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038034.1 2 - 3 340 97
          NR_038035.1 2 - 3 340 97
          GenBank mRNA AK029596.1 - 2 - 3 341 97
          EU523860.1 - 1 - 2 92 97
          EU523861.1 - 2 - 3 186 97
          EU523864.1 - 2 - 3 186 97
          EU523865.1 - 2 - 3 186 97
          EU523868.1 - 2 - 3 186 97
          EU523869.1 - 2 - 3 186 97
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY714652.1 - 2 - 3 339 Chr. 10 - 18802997 - 18831928 [-] on Build GRCm38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          32926
          Gm32926 Anti-sense Chr. 10 - 18783575 - 18837997 on NCBI Build GRCm38
          Downstream PERP,
          TP53 apoptosis effector
          Perp 1110017A08Rik,
          KCP1,
          KRTCAP1,
          PIGPC1,
          THW
          26.2kb Anti-sense Chr. 10 - 18845071 - 18857073 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001811.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107631 - -
          Whole Mouse Genome G4122A A_52_P100231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08236789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08298057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08802919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08675372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08942622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08404263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08404258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08942623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08942621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26977562 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1439402_PM_at - -
          Mouse Genome 430 2.0 Array 1439402_PM_at - -
          Mouse Genome 430 2.0 Array 1439402_at - -
          Affymetrix Mouse Clariom S Array PSR1000013201.mm.1 TC1000001811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013201.mm.1 NR_038034.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013201.mm.1 NR_038034.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013202.mm.1 TC1000001811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013202.mm.1 NR_038034.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013202.mm.1 NR_038034.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013193.mm.1 TC1000001811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013193.mm.1 NR_038035.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013193.mm.1 NR_038035.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013200.mm.1 TC1000001811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013200.mm.1 NR_038034.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013200.mm.1 NR_038034.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013196.mm.1 TC1000001811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013196.mm.1 NR_038035.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013196.mm.1 NR_038035.1 Refseq
          Mouse Expression Set 430 1446239_PM_at - -
          Mouse Expression Set 430 1439402_at - -
          Mouse Expression Set 430 1439402_PM_at - -
          Mouse Expression Set 430 1446239_at - -
          Murine Genome U74v2 Set 166336_PM_at - -
          Murine Genome U74v2 Set 166336_at - -
          Affymetrix HT Mouse Genome 430B Array 1439402_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439402_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103940332 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0