Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01191057_m1
          See other D030045P18RIK GE Assays ›
          Gene Symbol:
          D030045P18Rik
          Gene Name:
          RIKEN cDNA D030045P18 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 45807704 - 45852018 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01191057_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D030045P18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040624.1 2 - 3 503 85
          GenBank mRNA AK080062.1 - 2 - 3 504 85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB637151.1 - 2 - 3 504 Chr. 10 - 45807704 - 45850736 [+] on Build GRCm38 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream HECT domain and ankyrin repeat containing,
          E3 ubiquitin protein ligase 1
          Hace1 1700042J16Rik,
          A730034A22Rik,
          BC025474
          -116.4kb Sense Chr. 10 - 45577571 - 45712345 on NCBI Build GRCm38
          Downstream proteasome (prosome,
          macropain) subunit,
          alpha type 6 pseudogene
          C130030K03Rik 3.2mb Anti-sense Chr. 10 - 49093681 - 49095843 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000432.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2248335 AK050962.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 861514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808455 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41341 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331266 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916312 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741985 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96218 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177689 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02117390 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07048676 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02117370 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07048634 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07048613 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110278 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02117390 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07048613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08493557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27112585 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000002932.mm.1 TC1000000432.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000002932.mm.1 NR_040624.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000002932.mm.1 NR_040624.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 459079 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104150059 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0