Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01191145_m1
          See other PLA2G3 GE Assays ›
          Gene Symbol:
          Pla2g3
          Gene Name:
          phospholipase A2, group III
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 3487596 - 3499346 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01191145_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pla2g3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172791.2 NP_766379.2 6 - 7 1425 96
          GenBank mRNA AK033572.1 - 6 - 7 1369 96
          AK046215.1 - 3 - 4 1539 96
          BC079556.1 - 6 - 7 1425 30360278 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000014.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pla2g3

          Gene Ontology Categories:

          Function(s) Process(es)

          acrosome assembly
          sperm axoneme assembly
          lipoxygenase pathway
          mast cell degranulation
          phosphatidylcholine metabolic process
          cell development
          cilium assembly
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150851 AK081075.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150851 AK165718.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043033 BC079556.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043033 NM_172791.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043039 BC079556.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043039 AK033572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043039 AK046215.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043039 NM_172791.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815170 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68703 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133805 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153777 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150851 AK081075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150851 AK165718.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043039 BC079556.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043039 AK033572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043039 AK046215.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043039 NM_172791.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043033 BC079556.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043033 NM_172791.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402868 BC079556.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402868 AK033572.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402868 AK046215.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402868 NM_172791.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552261 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133805 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07238269 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153777 - -
          Whole Mouse Genome G4122A A_52_P98581 AK081075.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402868 BC079556.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402868 AK033572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402868 AK046215.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402868 NM_172791.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101474 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133805 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153777 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09695751 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09695756 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09695750 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09695753 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09695744 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09695743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08998843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09579734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09695750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09079809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09519773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09259805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09259795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09057326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09695756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09482504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09205297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09602351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09695746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09057321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09482505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08998845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09519775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27552267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09057327 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373764 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373762 - -
          Affymetrix HT MG-430 PM Array Plate 1456231_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373756 - -
          Mouse Genome 430 2.0 Array 1456231_at - -
          Mouse Genome 430 2.0 Array 1456231_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100000233.mm.1 TC1100000014.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000233.mm.1 NM_172791.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000233.mm.1 NM_172791.2 Refseq
          Mouse Expression Set 430 1456231_at - -
          Mouse Expression Set 430 1456231_PM_at - -
          Murine Genome U74v2 Set 166577_i_at - -
          Murine Genome U74v2 Set 166577_PM_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1456231_at - -
          Affymetrix HT Mouse Genome 430B Array 1456231_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102650113 - -
          Illumina mouse-6 v1.0 expression beadchip 104480725 - -
          Illumina mouse-6 v1.0 expression beadchip 5130739 - -
          Illumina mouse-6 v1.0 expression beadchip 102650113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005130739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240081 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215042 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240081 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229657 - -
          mouse-6 v1.1 (Illumina) scl41905.8.1_18-S - -
          Illumina Sentrix mouse-6.v1 bead array 5130739 - -
          Illumina Sentrix mouse-6.v1 bead array 102650113 - -
          Illumina Sentrix mouse-6.v1 bead array 104480725 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41905.8.1_18-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104480725 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5130739 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102650113 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1240081 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2565738 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240081 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215042 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0