Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01191785_m1
          See other MMACHC GE Assays ›
          Gene Symbol:
          Mmachc
          Gene Name:
          methylmalonic aciduria cblC type, with homocystinuria
          Gene Aliases:
          1810037K07Rik
          Chromosome Location:
          Chr.4: 116702434 - 116708385 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01191785_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mmachc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025962.3 NP_080238.2 3 - 4 496 112
          GenBank mRNA AK007725.1 - 3 - 4 431 112
          AK012761.1 - 3 - 4 433 112
          AK084194.1 - 3 - 4 447 112
          AK213975.1 - 2 - 3 197 112
          BB614283.1 - 3 - 4 497 112
          BC054756.1 - 3 - 4 435 6411797 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003349.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mmachc

          Gene Ontology Categories:

          Function(s) Process(es)

          cobalamin metabolic process
          molecular_function
          cobalamin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388298 AK084194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388298 BC054756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388298 NM_025962.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388298 AK007725.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388298 AK084194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388298 BC054756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388298 NM_025962.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388298 AK007725.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318640 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141513 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388298 AK084194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388298 BC054756.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388298 NM_025962.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388298 AK007725.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388298 AK084194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388298 BC054756.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388298 NM_025962.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388298 AK007725.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230536 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141513 - -
          Whole Mouse Genome G4122A A_51_P388298 AK084194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388298 BC054756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388298 NM_025962.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388298 AK007725.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388298 AK084194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388298 BC054756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388298 NM_025962.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388298 AK007725.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135943 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141513 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03879485 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03879481 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03879473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03879486 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03879475 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03879482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03972377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03966899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03851717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03966898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03208539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03725056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230537 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10515365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515367 - -
          Affymetrix HT MG-430 PM Array Plate 1418915_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10515363 - -
          Mouse Genome 430 2.0 Array 1418915_PM_at - -
          Mouse Genome 430 2.0 Array 1418915_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67096_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67096_at - -
          Affymetrix Mouse Clariom S Array PSR0400026770.mm.1 TC0400003349.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026770.mm.1 NM_025962.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026770.mm.1 NM_025962.3 Refseq
          Mouse Expression Set 430 1418915_PM_at - -
          Mouse Expression Set 430 1418915_at - -
          Mouse Genome 430A 2.0 Array 1418915_at - -
          Mouse Genome 430A 2.0 Array 1418915_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418915_at - -
          Affymetrix HT Mouse Genome 430A Array 1418915_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 753573 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6380176 - -
          Illumina mouse-6 v1.0 expression beadchip 6380176 - -
          Illumina mouse-6 v1.0 expression beadchip 102320731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380176 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221958 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2446992 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221958 - -
          mouse-6 v1.1 (Illumina) scl23949.4_43-S - -
          mouse-6 v1.1 (Illumina) scl0067096.2_233-S - -
          Illumina Sentrix mouse-6.v1 bead array 102320731 - -
          Illumina Sentrix mouse-6.v1 bead array 6380176 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067096.2_233-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102320731 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380176 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2446990 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2446990 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0067096.2_233-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0