Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01192495_m1
          See other STX11 GE Assays ›
          Gene Symbol:
          Stx11
          Gene Name:
          syntaxin 11
          Gene Aliases:
          5830405C08Rik
          Chromosome Location:
          Chr.10: 12939983 - 12964500 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01192495_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stx11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029075.1 NP_083351.1 1 - 2 263 54
          GenBank mRNA AK017897.1 - 1 - 2 86 54
          AK153408.1 - 1 - 2 238 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001761.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK017897.1, AK153408.1, NM_029075.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          T cell mediated cytotoxicity
          transport
          intracellular protein transport
          protein transport
          vesicle-mediated transport
          synaptic vesicle fusion to presynaptic active zone membrane
          natural killer cell mediated cytotoxicity
          neutrophil degranulation
          cytotoxic T cell degranulation
          natural killer cell degranulation
          vesicle docking
          SNARE binding
          SNAP receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948304 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948305 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948307 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948224 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948306 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948303 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948223 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513568 BC118516.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513568 AK153408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513568 NM_001163591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513568 NM_029075.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513568 AK017897.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513568 BC118516.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513568 AK153408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513568 NM_001163591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513568 NM_029075.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513568 AK017897.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494162 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137956 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 117010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465129 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9266 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148662 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156596 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153348 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513568 BC118516.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513568 AK153408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513568 NM_001163591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513568 NM_029075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513568 AK017897.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513568 BC118516.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513568 AK153408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513568 NM_001163591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513568 NM_029075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513568 AK017897.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P513568 BC118516.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P513568 AK153408.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P513568 NM_001163591.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P513568 NM_029075.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P513568 AK017897.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P513568 BC118516.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P513568 AK153408.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P513568 NM_001163591.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P513568 NM_029075.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P513568 AK017897.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156596 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06979177 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153348 - -
          Whole Mouse Genome G4122A A_51_P513568 BC118516.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513568 AK153408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513568 NM_001163591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513568 NM_029075.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513568 AK017897.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513568 BC118516.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513568 AK153408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513568 NM_001163591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513568 NM_029075.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513568 AK017897.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153348 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156596 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948303 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948306 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948304 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948223 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948305 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948307 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26948224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08398385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08715092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08463935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08485847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08511247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26948356 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10367921 - -
          Affymetrix HT MG-430 PM Array Plate 1457780_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453228_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10367919 - -
          Mouse Genome 430 2.0 Array 1453228_PM_at - -
          Mouse Genome 430 2.0 Array 1453228_at - -
          Mouse Genome 430 2.0 Array 1457780_PM_at - -
          Mouse Genome 430 2.0 Array 1457780_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74732_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74732_at - -
          Affymetrix Mouse Clariom S Array PSR1000012834.mm.1 TC1000001761.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000012834.mm.1 NM_029075.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012834.mm.1 NM_029075.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012837.mm.1 TC1000001761.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000012837.mm.1 NM_001163590.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012837.mm.1 NM_001163590.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012837.mm.1 NM_029075.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012837.mm.1 NM_029075.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012836.mm.1 TC1000001761.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000012836.mm.1 NM_029075.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012836.mm.1 NM_029075.1 Refseq
          Mouse Expression Set 430 1457780_PM_at - -
          Mouse Expression Set 430 1453228_at - -
          Mouse Expression Set 430 1457780_at - -
          Mouse Expression Set 430 1453228_PM_at - -
          Murine Genome U74v2 Set 134289_PM_r_at - -
          Murine Genome U74v2 Set 134041_at - -
          Murine Genome U74v2 Set 134289_r_at - -
          Murine Genome U74v2 Set 134041_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453228_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457780_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457780_at - -
          Affymetrix HT Mouse Genome 430B Array 1453228_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 330925 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6590184 - -
          MouseWG-6 v2 Expression BeadChip 3710528 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2734340 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2734340 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0