Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01192779_g1
          See other TEX38 GE Assays ›
          Gene Symbol:
          Tex38
          Gene Name:
          testis expressed 38
          Gene Aliases:
          4930544O15Rik, Theg4
          Chromosome Location:
          Chr.4: 115779831 - 115782759 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01192779_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tex38
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029196.1 NP_083472.1 1 - 2 53 57
          GenBank mRNA CF982527.1 - 1 - 2 53 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003323.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_029196.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of mitophagy in response to mitochondrial depolarization
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081690 FJ468007.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081690 NM_029196.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081690 AK016041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081690 BC119474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081690 BC152479.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287798 NM_029196.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287798 AK016041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287798 BC119474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287798 BC152479.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287798 NM_029196.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287798 AK016041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287798 BC119474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287798 BC152479.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287798 NM_029196.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287798 AK016041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287798 BC119474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287798 BC152479.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287798 NM_029196.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287798 AK016041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287798 BC119474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287798 BC152479.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081690 FJ468007.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081690 NM_029196.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081690 AK016041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081690 BC119474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081690 BC152479.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287802 NM_029196.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287802 AK016041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287802 BC119474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287802 BC152479.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287798 NM_029196.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287798 AK016041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287798 BC119474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287798 BC152479.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287798 NM_029196.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287798 AK016041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287798 BC119474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287798 BC152479.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111857 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10515212 - -
          Affymetrix HT MG-430 PM Array Plate 1430517_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10515211 - -
          Mouse Genome 430 2.0 Array 1430517_at - -
          Mouse Genome 430 2.0 Array 1430517_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75173_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75173_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400026446.mm.1 TC0400003323.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026446.mm.1 NM_029196.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026446.mm.1 NM_029196.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026449.mm.1 TC0400003323.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026449.mm.1 NM_029196.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026449.mm.1 NM_029196.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026447.mm.1 TC0400003323.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026447.mm.1 NM_029196.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026447.mm.1 NM_029196.1 Refseq
          Mouse Expression Set 430 1430517_at - -
          Mouse Expression Set 430 1430517_PM_at - -
          Murine Genome U74v2 Set 168773_at - -
          Murine Genome U74v2 Set 168773_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430517_at - -
          Affymetrix HT Mouse Genome 430B Array 1430517_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2759624 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0