Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01193458_g1
          See other NPRL3 GE Assays ›
          Gene Symbol:
          Nprl3
          Gene Name:
          nitrogen permease regulator-like 3
          Gene Aliases:
          Aag, CGTHBA, HS-26, HS-40, Mare, Phg, Prox1, m(alpha)RE
          Chromosome Location:
          Chr.11: 32231963 - 32267707 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01193458_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nprl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001284359.1 NP_001271288.1 10 - 11 1235 82
          NM_001284360.1 NP_001271289.1 8 - 9 1066 82
          NM_181569.3 NP_853547.1 9 - 10 1196 82
          NR_104306.1 10 - 11 1416 82
          GenBank mRNA AK036405.1 - 9 - 10 1105 82
          AK036633.1 - 8 - 9 1165 82
          AK039600.1 - 8 - 9 1042 82
          AK047190.1 - 10 - 11 1144 82
          AK081902.1 - 9 - 10 1107 82
          AK086378.1 - 1009 82
          AK090290.1 - 82
          AK154884.1 - 10 - 11 1368 82
          BC064471.1 - 9 - 10 1196 6334386 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002473.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nprl3

          Gene Ontology Categories:

          Function(s) Process(es)

          ventricular septum development
          aorta morphogenesis
          cardiac muscle tissue development
          roof of mouth development
          molecular_function
          GTPase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100023044.mm.1 TC1100002473.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100023044.mm.1 NM_001284359.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023044.mm.1 NM_001284360.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023044.mm.1 NM_181569.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023044.mm.1 NR_104306.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023046.mm.1 TC1100002473.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100023046.mm.1 NM_001284359.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023046.mm.1 NM_001284360.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023046.mm.1 NM_181569.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023046.mm.1 NR_104306.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023045.mm.1 TC1100002473.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100023045.mm.1 NM_001284359.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023045.mm.1 NM_001284360.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023045.mm.1 NM_181569.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023045.mm.1 NR_104306.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023047.mm.1 TC1100002473.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100023047.mm.1 NM_001284359.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023047.mm.1 NM_001284360.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023047.mm.1 NM_181569.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023047.mm.1 NR_104306.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0