Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01193768_m1
          See other SMLR1 GE Assays ›
          Gene Symbol:
          Smlr1
          Gene Name:
          small leucine-rich protein 1
          Gene Aliases:
          2010003K15Rik
          Chromosome Location:
          Chr.10: 25527943 - 25536272 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01193768_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smlr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195596.1 NP_001182525.1 1 - 2 313 66
          GenBank mRNA AK008073.1 - 1 - 2 311 66
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY708055.1 - 1 - 2 311 Chr. 10 - 25527956 - 25536267 [-] on Build GRCm38 66
          GenBankmRNA BY034367.1 - 1 - 2 314 Chr. 10 - 25528235 - 25536272 [-] on Build GRCm38 66
          GenBankmRNA CA478867.1 - 204 Chr. - - [-] on Build 66
          GenBankmRNA CA456672.1 - 175 Chr. - - [-] on Build 66

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 29571 Gm29571 Anti-sense Chr. 10 - 25536252 - 25703341 on NCBI Build GRCm38
          Downstream predicted gene,
          36908
          Gm36908 78.5kb Sense Chr. 10 - 25614752 - 25621965 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001905.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 104014 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P96847 NM_001195596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P96847 AK008073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P96847 NM_001195596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P96847 AK008073.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07007000 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27014937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27014930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27014927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27014929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27014919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27014934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27014920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27014932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08419229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27014939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27014924 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75606_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75606_at - -
          Affymetrix Mouse Clariom S Array PSR1000013732.mm.1 TC1000001905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013732.mm.1 NM_001195596.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013732.mm.1 NM_001195596.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013735.mm.1 TC1000001905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013735.mm.1 NM_001195596.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013735.mm.1 NM_001195596.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013734.mm.1 TC1000001905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013734.mm.1 NM_001195596.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013734.mm.1 NM_001195596.1 Refseq
          Murine Genome U74v2 Set 165935_PM_at - -
          Murine Genome U74v2 Set 165935_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 622074 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0