Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01194476_m1
          See other ANAPC5 GE Assays ›
          Gene Symbol:
          Anapc5
          Gene Name:
          anaphase-promoting complex subunit 5
          Gene Aliases:
          2510006G12Rik, AA408751, AA536819, AA986414, APC5, Anpc5
          Chromosome Location:
          Chr.5: 122787459 - 122821342 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01194476_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Anapc5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042491.2 NP_001035956.1 3 - 4 548 130
          NM_001289517.1 NP_001276446.1 3 - 4 548 130
          NM_001289518.1 NP_001276447.1 3 - 4 548 130
          NM_001289519.1 NP_001276448.1 3 - 4 548 130
          NM_001289520.1 NP_001276449.1 3 - 4 548 130
          NM_021505.3 NP_067480.2 3 - 4 548 130
          GenBank mRNA AB041593.1 - 3 - 4 491 130
          AK013978.1 - 3 - 4 472 130
          AK031432.1 - 3 - 4 473 130
          AK049719.1 - 3 - 4 502 130
          AK075969.1 - 3 - 4 487 130
          AK088327.1 - 3 - 4 511 130
          AK147951.1 - 3 - 4 487 130
          AK148679.1 - 3 - 4 503 130
          AK151314.1 - 3 - 4 494 130
          AK151905.1 - 3 - 4 494 130
          AK152528.1 - 3 - 4 464 130
          AK152981.1 - 3 - 4 494 130
          AK153373.1 - 3 - 4 494 130
          AK153422.1 - 3 - 4 494 130
          AK159713.1 - 3 - 4 487 130
          AK159916.1 - 3 - 4 487 130
          AK167561.1 - 3 - 4 487 130
          AK168135.1 - 3 - 4 472 130
          AK168737.1 - 3 - 4 488 130
          AK168814.1 - 3 - 4 487 130
          AK169196.1 - 3 - 4 511 130
          AK192754.1 - 3 - 4 244 130
          AK194108.1 - 3 - 4 213 130
          AK197886.1 - 3 - 4 220 130
          AK203216.1 - 2 - 3 197 130
          AK217317.1 - 3 - 4 226 130
          BC010339.1 - 3 - 4 464 2648173 130
          BC046566.1 - 3 - 4 527 2650790 130
          BC046804.1 - 3 - 4 485 6404923 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003220.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Anapc5

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          cell cycle
          cell division
          protein K11-linked ubiquitination
          protein phosphatase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500028817.mm.1 TC0500003220.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500028817.mm.1 NM_001042491.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500028817.mm.1 NM_001289517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500028817.mm.1 NM_001289518.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500028817.mm.1 NM_001289519.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500028817.mm.1 NM_001289520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500028817.mm.1 NM_021505.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0