Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01194986_g1
          See other CCDC110 GE Assays ›
          Gene Symbol:
          Ccdc110
          Gene Name:
          coiled-coil domain containing 110
          Gene Aliases:
          Gm172
          Chromosome Location:
          Chr.8: 45934649 - 45945230 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01194986_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc110
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033246.2 NP_001028418.1 6 - 7 2552 154
          GenBank mRNA AK132742.1 - 6 - 7 2553 154
          AK161248.1 - 6 - 7 2540 154

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000523.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P363316 AK132742.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P363316 AK161248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P363316 NM_001033246.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P363316 AK132742.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P363316 AK161248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P363316 NM_001033246.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959932 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782878 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 314563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36994 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P363316 AK132742.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P363316 AK161248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P363316 NM_001033246.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P363316 AK132742.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P363316 AK161248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P363316 NM_001033246.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25833372 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06498825 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01727497 - -
          Whole Mouse Genome G4122A A_52_P363316 AK132742.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P363316 AK161248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P363316 NM_001033246.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P363316 AK132742.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P363316 AK161248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P363316 NM_001033246.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01727497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25833438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07301341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07086667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07145047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07123695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06939197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06865777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06965009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06865776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25833415 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10571618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571615 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571613 - -
          Affymetrix HT MG-430 PM Array Plate 1458388_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10571612 - -
          Mouse Genome 430 2.0 Array 1458388_at - -
          Mouse Genome 430 2.0 Array 1458388_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 212392_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 212392_at - -
          Affymetrix Mouse Clariom S Array PSR0800003658.mm.1 TC0800000523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800003658.mm.1 NM_001033246.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800003658.mm.1 NM_001033246.2 Refseq
          Mouse Expression Set 430 1458388_at - -
          Mouse Expression Set 430 1458388_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458388_at - -
          Affymetrix HT Mouse Genome 430B Array 1458388_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 334440 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2823947 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2823947 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0